Skip to Content
Merck
All Photos(2)

Key Documents

19329

Supelco

Omeprazole

analytical standard

Synonym(s):

5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, Antra, Losec

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3O3S
CAS Number:
Molecular Weight:
345.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)c(OC)c3C

InChI

1S/C17H19N3O3S/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)

InChI key

SUBDBMMJDZJVOS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofang Huo et al.
Gut, 63(7), 1042-1052 (2013-09-21)
Oesophagitis might result from the effects of chemokines produced by oesophageal cells in response to gastro-oesophageal reflux, and not solely from the direct, caustic effects of refluxed gastric juice. Proton pump inhibitors (PPI) can block chemokine production through mechanisms independent
J Dent
Alimentary pharmacology & therapeutics, 17 Suppl 1, 5-9 (2003-03-05)
Plasma concentration measurements have confirmed that the advantageous hepatic metabolism of esomeprazole results in a greater delivery of acid suppressant to the systemic circulation, compared with an equal dose of omeprazole. Also, this superior delivery has been shown to cause
R P Myers et al.
The American journal of gastroenterology, 96(12), 3428-3431 (2002-01-05)
Omeprazole is a proton pump inhibitor that is used commonly in the treatment of acid-peptic disorders. Although omeprazole is generally well tolerated, serious adverse effects such as renal failure have been reported. Thus far, 17 cases of acute interstitial nephritis
Anju Nohria et al.
Journal of the American Heart Association, 3(1), e000609-e000609 (2014-01-07)
Inflammation is fundamental to the development of atherosclerosis. We examined the effect of anti-inflammatory doses of salicylate on endothelium-dependent vasodilation, a biomarker of cardiovascular risk, in a broad range of subjects. We performed a randomized, double-blind, placebo-controlled crossover trial evaluating
A proton-pump inhibitor expedition: the case histories of omeprazole and esomeprazole.
Lars Olbe et al.
Nature reviews. Drug discovery, 2(2), 132-139 (2003-02-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service