Skip to Content
Merck
All Photos(1)

Key Documents

45548

Supelco

Lindane

PESTANAL®, analytical standard

Synonym(s):

γ-BHC, 1α,2α,3β,4α,5α,6β-Hexachlorocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Beilstein:
1907337
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

113-115 °C (lit.)

solubility

H2O: 8.35 g/L at 25 °C

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cl[C@H]1[C@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+

InChI key

JLYXXMFPNIAWKQ-GNIYUCBRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lindane is an organochlorine pesticide, widely employed in agriculture and in the pharmaceutical treatment of scabies and lice.

Application

Lindane may be used as an analytical reference standard for the quantification of the analyte in environmental samples using isotope dilution method with gas chromatography/mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Design and fabrication of molecularly imprinted polymer-based potentiometric sensor from the surface modified multiwalled carbon nanotube for the determination of lindane (gamma-hexachlorocyclohexane), an organochlorine pesticide
Anirudhan.ST and Alexander S
Biosensors And Bioelectronics, 64, 586-593 (2015)
Determination of atrazine, lindane, pentachlorophenol, and diazinon in water and soil by isotope dilution gas chromatography/mass spectrometry.
Lopez-Avila V, et al.
Analytical Chemistry, 57(14), 2797-2801 (1985)
L A Videla et al.
Free radical biology & medicine, 9(2), 169-179 (1990-01-01)
The development of an oxidative stress condition in the liver by lindane intoxication is discussed as a possible hepatotoxic mechanism of the insecticide. Lindane is metabolized by liver microsomal enzymes to a variety of metabolites, which are susceptible of conjugation
Microbial degradation of the pesticide lindane (gamma-hexachlorocyclohexane).
B K Singh et al.
Advances in applied microbiology, 47, 269-298 (2003-07-25)
Yuji Nagata et al.
Applied microbiology and biotechnology, 76(4), 741-752 (2007-07-20)
gamma-Hexachlorocyclohexane (gamma-HCH, also called gamma-BHC and lindane) is a halogenated organic insecticide that causes serious environmental problems. The aerobic degradation pathway of gamma-HCH was extensively revealed in bacterial strain Sphingobium japonicum (formerly Sphingomonas paucimobilis) UT26. gamma-HCH is transformed to 2,5-dichlorohydroquinone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service