Merck
  • Cognitive changes after saline or plasmalyte infusion in healthy volunteers: a multiple blinded, randomized, cross-over trial.
Cognitive changes after saline or plasmalyte infusion in healthy volunteers: a multiple blinded, randomized, cross-over trial.
Anesthesiology (2013-04-20)
David A Story, Lucy Lees, Laurence Weinberg, Soon-Yee Teoh, Katherine J Lee, Sarah Velissaris, Rinaldo Bellomo, Sarah J Wilson
ABSTRACT
In an incidental finding, during a study of plasma chemistry after crystalloid infusion, participants reported subjective cognitive changes, particularly slower thinking, after saline but not Hartmann's (Ringer's lactate) solution. The authors tested the hypothesis that saline infusion would produce greater adverse cognitive changes than Plasmalyte infusion. The authors conducted a randomized, cross-over, multiple blinded study of healthy adult volunteers. On separate days, participants received 30 ml/kg over 1 h of either 0.9% saline or Plasmalyte with the order randomly allocated. Plasma chemistry was tested on venous samples. As part of a battery of cognitive tests our primary endpoint was the reaction time index after infusion. The authors studied 25 participants. Plasma chloride was greater after saline than after Plasmalyte: mean difference 5.4 mM (95% CI, 4.1-6.6 mM; P < 0.001). Saline was also associated with greater metabolic acidosis: base-excess 2.5 mM more negative (95% CI, 1.9-3.0 mM more negative; P < 0.001). There was no evidence of a difference in the reaction time index between the two interventions: mean reaction time index 394 ms (SD, 72) after saline versus 385 ms (SD, 55) after Plasmalyte. Difference: saline 9 ms slower (95% CI, 30 ms slower to 12 ms faster; P = 0.39). There were minimal differences in the other cognitive and mood tests. Despite expected differences in plasma chemistry, the authors found that measures of cognition did not differ after infusions of Plasmalyte or saline.
Product Description
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~0.025 M in H2O
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, 2 M in H2O
Sigma-Aldrich
Potassium chloride, ≥99.0%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications