Skip to Content
Merck
All Photos(1)

Documents

60126

Sigma-Aldrich

Potassium chloride

tested according to Ph. Eur.

Synonym(s):

Kalii chloridum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanne Sørup Tastesen et al.
Amino acids, 46(7), 1659-1671 (2014-03-25)
High-protein diets induce alterations in metabolism that may prevent diet-induced obesity. However, little is known as to whether different protein sources consumed at normal levels may affect diet-induced obesity and associated co-morbidities. We fed obesity-prone male C57BL/6J mice high-fat, high-sucrose
Gregory Saia et al.
Journal of neurochemistry, 129(4), 743-752 (2014-01-31)
The regulation of transcription factor function in response to neuronal activity is important for development and function of the nervous system. The transcription factor Sp4 regulates the developmental patterning of dendrites, contributes to complex processes including learning and memory, and
Sandra S Wise et al.
Toxicology and applied pharmacology, 279(2), 113-118 (2014-06-22)
Sea turtles are a charismatic and ancient ocean species and can serve as key indicators for ocean ecosystems, including coral reefs and sea grass beds as well as coastal beaches. Genotoxicity studies in the species are absent, limiting our understanding
David A Story et al.
Anesthesiology, 119(3), 569-575 (2013-04-20)
In an incidental finding, during a study of plasma chemistry after crystalloid infusion, participants reported subjective cognitive changes, particularly slower thinking, after saline but not Hartmann's (Ringer's lactate) solution. The authors tested the hypothesis that saline infusion would produce greater
Siti Shahara Zulfakar et al.
Food microbiology, 34(2), 369-375 (2013-04-02)
Microbial contamination of carcass surfaces occurs during slaughter and post-slaughter processing steps, therefore interventions are needed to enhance meat safety and quality. Although many studies have been done at the macro-level, little is known about specific processes that influence bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service