Skip to Content
Merck
All Photos(1)

Key Documents

12636

Sigma-Aldrich

Potassium chloride

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals
Pricing and availability is not currently available.

grade

puriss.

Quality Level

Assay

99-100.5% (AT)

form

powder or crystals

quality

meets analytical specification of Ph. Eur., BP, USP, FCC, E508

impurities

acidity or alkalinity, complies
organic volatile impurities, complies (GC)
residual solvents, complies
≤0.0001% Al
≤0.0005% heavy metals (as Pb)
≤0.003% free acid (as HCl)
≤0.005% free alkali (as KOH)
≤0.02% Mg, alkaline earth metals (as Ca)

loss

≤0.5% loss on drying, 105 °C, 3 h

pH

7

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤1000 mg/kg
iodide (I-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.5 mg/kg
Ba:, in accordance
Ca: ≤50 mg/kg
Cd: ≤1 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
Mg: ≤50 mg/kg
Na: ≤700 mg/kg
Pb: ≤5 mg/kg
Zn: ≤20 mg/kg

suitability

complies for appearance of solution

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium chloride, an alkali metal halide, is a colorless crystalline salt. Its solubility in water is directly proportional to increase in temperature. It can be prepared from potassium hydroxide, via neutralization reaction with HCl.[1] Calorimetric estimation of the enthalpy of solution of potassium chloride in water has been described.[2]

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.
Potassium chloride may be used in the preparation of fertilizer, potassium hydroxide, potassium carbonate and other potassium slats.[1] It may be used in the calorimetric determination of enthalpy and heat capacity values during the deprotonation (loss of proton H+) of buffers. From these values temperature dependence of the pH of these buffers was calculated.[3]
Potassium chloride can be used as a fertilizer and as a regenerant to soften water.[4] It has been used a buffer for the separation of major purine metabolites in the heart, using reverse phase ion pair chromatography(RPIPC).[5]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

McEachern R, et al.
Water Softening with Potassium Chloride: Process, Health, and Environmental Benefits (2009)
Determination of sixteen nucleotides, nucleosides and bases using high-performance liquid chromatography and its application to the study of purine metabolism in hearts for transplantation
Smolenski,T.R, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 527, 414-420 (1990)
H Fukada et al.
Proteins, 33(2), 159-166 (1998-10-21)
Enthalpy and heat capacity changes for the deprotonation of 18 buffers were calorimetrically determined in 0.1 M potassium chloride at temperatures ranging from 5 to 45 degrees C. The values of the dissociation constant were also determined by means of
The use of potassium chloride and tris (hydroxymethyl) aminomethane as standard substances for solution calorimetry.
Rychly R and Pekarek V.
The Journal of Chemical Thermodynamics, 9(4), 391-396 (1977)
Masatoshi Takagaki et al.
Experimental neurology, 252, 12-17 (2013-11-20)
Sedatives in the neurointensive care unit can strongly influence patients' risks of developing secondary brain damage. In particular, isoflurane, a volatile anesthetic, has been recently re-introduced to the neurointensive care unit, and first clinical studies suggest beneficial effects due to

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service