Merck
  • New insights into the initial stages of Ta oxide nanotube formation on polycrystalline Ta electrodes.
New insights into the initial stages of Ta oxide nanotube formation on polycrystalline Ta electrodes.
Nanoscale (2013-01-23)
Hany A El-Sayed, Corie A Horwood, Anusha D Abhayawardhana, Viola I Birss
ABSTRACT
Ta oxide nanotubes (NTs) were formed by the anodization of Ta at 15 V in a solution of concentrated sulfuric acid containing 0.8-1.0 M hydrofluoric acid. To study the initial stages of NT formation, FESEM images of samples anodized for very short times were obtained. The results contradict the existing explanation of the current-time data collected during anodization, which has persisted in the literature for more than two decades. In addition to providing a first-time morphological study of Ta oxide NT formation at very early stages of anodization, we also propose a new interpretation of the i-t response, showing that pores are already present in the first few milliseconds of anodization and that NTs are formed well before present models predict. This behaviour may also extend to the anodization of other valve metals, such as Al, Ti, Zr, W, and Nb.
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis