Skip to Content
Merck
All Photos(1)

Key Documents

262862

Sigma-Aldrich

Tantalum

wire, diam. 1.0 mm, ≥99.9% trace metals basis

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
EC Number:
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9% trace metals basis

form

wire

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

diam.

1.0 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Quantity

13 g = 1 m; 65 g = 5 m

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frédéric Clarençon et al.
Journal of neurosurgery. Spine, 18(6), 606-610 (2013-04-23)
Presurgical devascularization of hypervascular spinal metastases has been shown to be effective in preventing major blood loss during open surgery. Most often, embolization can be performed using polyvinyl alcohol (PVA) microparticles. However, in some cases, the close relationship between the
A M Ashmore et al.
Journal of biomechanics, 45(16), 2931-2934 (2012-10-13)
Interest in soft tissue RadioStereometric Analysis (RSA) is rising. Previous studies have attempted to use this technique to analyse anterior cruciate ligament (ACL) graft constructs, and more recently, the movement of soft tissue as a precursor to RSA follow up
Biomimetic oxygen activation by MoS2/Ta3N5 nanocomposites for selective aerobic oxidation.
Qingsheng Gao et al.
Angewandte Chemie (International ed. in English), 51(47), 11740-11744 (2012-10-17)
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
Hamid Shokoohi et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 137-139 (2013-02-26)
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service