Skip to Content
Merck
All Photos(1)

Key Documents

262889

Sigma-Aldrich

Tantalum

foil, thickness 1.0 mm, ≥99.9% trace metals basis

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
EC Number:
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9% trace metals basis

form

foil

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

thickness

1.0 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Related Categories

Quantity

10.4 g = 25 × 25 mm; 41.6 g = 50 × 50 mm

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nag et al.
Journal of the mechanical behavior of biomedical materials, 16, 21-28 (2012-11-10)
Microstructure and mechanical properties of laser deposited complex quaternary Ti-34Nb-7Zr-7Ta (all wt%), an orthopedic load-bearing implant alloy, has been investigated in detail in both as-deposited as well as heat-treated (β-solutionized and quenched) conditions. The difference in stress-strain behavior of this
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization
Hany A El-Sayed et al.
Nanoscale, 5(4), 1494-1498 (2013-01-23)
Ta oxide nanotubes (NTs) were formed by the anodization of Ta at 15 V in a solution of concentrated sulfuric acid containing 0.8-1.0 M hydrofluoric acid. To study the initial stages of NT formation, FESEM images of samples anodized for
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
Hamid Shokoohi et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 137-139 (2013-02-26)
Re: Fernandez-Fairen M, Murcia A, Torres A, et al. Is anterior cervical fusion with a porous tantalum implant a cost-effective method to treat cervical disc disease with radiculopathy? Spine 2012; 37:1734–41.
Robert A Beatty
Spine, 38(4), 369-369 (2012-10-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service