Skip to Content
Merck
All Photos(1)

Key Documents

357251

Sigma-Aldrich

Tantalum

foil, thickness 0.5 mm, ≥99.9% trace metals basis

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
EC Number:
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9% trace metals basis

form

foil

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

thickness

0.5 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Related Categories

Quantity

20.8 g = 50 × 50 mm; 83.2 g = 100 × 100 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongyuan Guo et al.
International journal of molecular medicine, 31(3), 689-697 (2013-01-23)
Titanium (Ti) alloys of the β-type are highly attractive metallic materials for biomedical applications due to their low elastic modulus, high corrosion resistance and notable biocompatibility. A new β-type Ti35Nb2Ta3Zr alloy with a low Young's modulus of approximately 48 GPa was previously
Biomimetic oxygen activation by MoS2/Ta3N5 nanocomposites for selective aerobic oxidation.
Qingsheng Gao et al.
Angewandte Chemie (International ed. in English), 51(47), 11740-11744 (2012-10-17)
Hany A El-Sayed et al.
Nanoscale, 5(4), 1494-1498 (2013-01-23)
Ta oxide nanotubes (NTs) were formed by the anodization of Ta at 15 V in a solution of concentrated sulfuric acid containing 0.8-1.0 M hydrofluoric acid. To study the initial stages of NT formation, FESEM images of samples anodized for
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
Hamid Shokoohi et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 137-139 (2013-02-26)
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service