Skip to Content
Merck
All Photos(1)

Key Documents

IRMM526C

Niobium

IRMM®, certified reference material, 0.5 mm wire

Synonym(s):

Columbium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

resistivity

13-16 μΩ-cm, 20°C

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

application(s)

general analytical

format

matrix material

SMILES string

[Nb]

InChI

1S/Nb

InChI key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM526C

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kulbir Kaur Ghuman et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085501-085501 (2013-01-25)
A systematic study of electronic structure and band gap states is conducted to analyze the monodoping and charge compensated codoping of rutile TiO(2) with Rh and Nb, using the DFT + U approach. Doping of rutile TiO(2) with Rh atoms induces hybridized
Evgeniy Papulovskiy et al.
Physical chemistry chemical physics : PCCP, 15(14), 5115-5131 (2013-03-02)
Ab initio DFT calculations of (93)Nb NMR parameters using the NMR-CASTEP code were performed for a series of over fifty individual niobates, and a good agreement has been found with experimental NMR parameters. New experimental and calculated (93)Nb NMR data
Anjan Soumyanarayanan et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(5), 1623-1627 (2013-01-16)
The competition between proximate electronic phases produces a complex phenomenology in strongly correlated systems. In particular, fluctuations associated with periodic charge or spin modulations, known as density waves, may lead to exotic superconductivity in several correlated materials. However, density waves
Huaxiang Yi et al.
Optics express, 20(25), 27562-27568 (2012-12-25)
We demonstrate error-free 80km transmission by a silicon carrier-depletion Mach-Zehnder modulator at 10Gbps and the power penalty is as low as 1.15dB. The devices were evaluated through the bit-error-rate characterizations under the system-level analysis. The silicon Mach-Zehnder modulator was also
Jun Deng et al.
Optics express, 21(3), 3243-3252 (2013-03-14)
In this paper the Fano resonance in a free-standing LiNbO(3) photonic crystal slab is demonstrated. We present a numerical analysis and experimental measurements with free space illumination where the dependence of slab thickness, radius of air holes and lattice types

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service