Merck
  • In-vitro Strain and Modulus Measurements in Porcine Cervical Lymph Nodes.
In-vitro Strain and Modulus Measurements in Porcine Cervical Lymph Nodes.
The open biomedical engineering journal (2011-06-07)
Queeny Wing-Han Yuen, Yong-Ping Zheng, Yan-Ping Huang, Jun-Feng He, James Chung-Wai Cheung, Michael Ying
ABSTRACT
Cervical lymph nodes are common sites of metastatic involvement in head and neck cancers. These lymph nodes are superficially located and palpation is a common practice for assessing nodal hardness and staging cancer which is, however, too subjective and with limited accuracy. In this study, the mechanical properties of pig lymph node tissues were investigated using ultrasound elastography and indentation test. Lymph nodes were excised from fresh pork pieces and embedded in an agar-gelatin phantom for strain imaging by elastography. A strain ratio reflecting the strain contrast of lymph node over agar-gelatin phantom was used to assess the elasticity of the lymph node. A cutting device was then custom-designed to slice the phantom into uniform slices for indentation test. The measurements revealed that there were significant differences in both the strain ratio and Young's modulus between the peripheral and middle regions of the lymph nodes (both p < 0.05); however, the results appeared contradictory. Correlation between the results of the two measurements (modulus ratio vs. inversed strain ratio) showed their association was moderate for both the peripheral and middle regions (R(2) = 0.437 and 0.424 respectively). As the tests were only performed on normal lymph nodes, comparison in stiffness between healthy and abnormal lymph nodes could not be made. Future studies should be conducted to quantify the stiffness change in abnormal lymph nodes.
Product Description
Millipore
Agar, suitable for microbiology
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology