Skip to Content
Merck
All Photos(2)

Key Documents

56763

Millipore

Agar

high purity, low ionic content, low gel strenght, suitable for microbiology

Synonym(s):

Agar-agar, Gum agar, Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

ign. residue

≤4.0%

loss

≤10% loss on drying

turbidity

≤8 NTU (after autoclaving)

mp

85-95 °C

transition temp

gel point 34-36 °C

gel strength

≥800 g/cm2

anion traces

sulfate (SO42-): ≤2.75%

cation traces

Ca: ≤500 mg/kg
Mg: ≤200 mg/kg

application(s)

microbiology

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Agar is of high purity, low ionic content, and low gel strength widely used in microbiology as a solidifying agent in microbial culture media. It is a polysaccharide derived from seaweed and is primarily composed of agarose. Agar is a mostly inert substance, which makes it an ideal material for microbial culture media because it doesn’t interfere with the growth of microorganisms. Agar can solidify media, allowing microorganisms to grow in a solid environment, which facilitates the formation of distinct colonies and selective isolation of specific microorganisms. Agar is also used in the preparation of selective and differential media, which can provide additional nutrients or other chemical substances to promote or inhibit the growth of specific microorganisms selectively. Agar is sterilized by autoclaving and then poured into Petri dishes, where it can cool and solidify, forming the surface for microbial growth.

Application

Agar is commonly used as a solidifying agent in many microbial culture media for the growth and identification of bacteria, yeasts and molds from various pharmaceutical, environmental, food and beverage samples.

Analysis Note

toxic substances: absence

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Arnaud Dechesne et al.
Applied and environmental microbiology, 74(16), 5195-5200 (2008-07-01)
Water is arguably the most important constituent of microbial microhabitats due to its control of physical and physiological processes critical to microbial activity. In natural environments, bacteria often live on unsaturated surfaces, in thin (micrometric) liquid films. Nevertheless, no experimental
Queeny Wing-Han Yuen et al.
The open biomedical engineering journal, 5, 39-46 (2011-06-07)
Cervical lymph nodes are common sites of metastatic involvement in head and neck cancers. These lymph nodes are superficially located and palpation is a common practice for assessing nodal hardness and staging cancer which is, however, too subjective and with
Jacky M Tiang et al.
PloS one, 6(2), e17031-e17031 (2011-02-25)
Arylamine N-acetyltransferase-1 (NAT1) is an enzyme that catalyzes the biotransformation of arylamine and hydrazine substrates. It also has a role in the catabolism of the folate metabolite p-aminobenzoyl glutamate. Recent bioinformatics studies have correlated NAT1 expression with various cancer subtypes.
GunNam Na et al.
The Journal of biological chemistry, 286(47), 40423-40432 (2011-09-21)
When growing in its native habitat, Thlaspi goesingense can hyperaccumulate 1.2% of its shoot dry weight as nickel. We reported previously that both constitutively elevated activity of serine acetyltransferase (SAT) and concentration of glutathione (GSH) are involved in the ability
Raphael H Matsunaga et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 5722-5725 (2013-02-01)
In this study, we presented an experiment to obtain the thermal relaxation time which is necessary to model heat conduction by the hyperbolic heat equation. This experiment was evaluated by finite element simulation to acquire reliably this parameter for biological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service