Skip to Content
Merck
All Photos(1)

Key Documents

48723

Sigma-Aldrich

Gelatin

tested according to Ph. Eur.

Synonym(s):

Gelatina

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Won Jai Lee et al.
Journal of Korean medical science, 26(8), 1081-1086 (2011-08-24)
Keloids are pathologic proliferations of the dermal layer of the skin resulting from excessive collagen production and deposition. Hepatocyte growth factor (HGF) increases the expression of matrix metalloproteinase (MMP)-1 and suppresses collagen synthesis to modulate extracellular matrix turnover. To investigate
Gloria Mosquera et al.
The Plant cell, 21(4), 1273-1290 (2009-04-10)
Biotrophic invasive hyphae (IH) of the blast fungus Magnaporthe oryzae secrete effectors to alter host defenses and cellular processes as they successively invade living rice (Oryza sativa) cells. However, few blast effectors have been identified. Indeed, understanding fungal and rice
Xiufeng Pang et al.
Cancer research, 69(2), 518-525 (2009-01-17)
Morelloflavone, a biflavonoid extracted from Garcinia dulcis, has shown antioxidative, antiviral, and anti-inflammatory properties. However, the function and the mechanism of this compound in cancer treatment and tumor angiogenesis have not been elucidated to date. In this study, we postulated
Gang Hu et al.
PloS one, 6(8), e23421-e23421 (2011-08-23)
Magnetoacoustic tomography with magnetic induction (MAT-MI) is an emerging approach for noninvasively imaging electrical impedance properties of biological tissues. The MAT-MI imaging system measures ultrasound waves generated by the Lorentz force, having been induced by magnetic stimulation, which is related
Wenlan Liu et al.
Journal of neurochemistry, 107(5), 1196-1205 (2008-09-13)
Matrix metalloproteinase-9 (MMP-9) and NADPH oxidase contribute to blood-brain barrier (BBB) disruption after ischemic stroke. We have previously shown that normobaric hyperoxia (NBO) treatment reduces MMP-9 and oxygen free radical generation in ischemic brain. In this study, we tested the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service