Skip to Content
Merck
All Photos(3)

Key Documents

A4550

Sigma-Aldrich

Agar

Type A, suitable for plant cell culture

Synonym(s):

Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
eCl@ss:
42040102
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

algae (Rhodophyceae)

Quality Level

type

Type A

form

powder

technique(s)

cell culture | plant: suitable

transition temp

transition temp 32-38 °C (1.5%)

gel strength

>650 g/cm2

application(s)

agriculture

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Agar (Type A) is extracted from agarocytes of the red alga Rhodophyceae. It is a polysaccharide complex, consisting of a neutral gelling fraction, agarose, and a non-gelling fraction, agaropectin. Type-A agar forms a thermoreversible gel[1], and is most suitable for plant cell culture.

Application

Type A agar has been used:
  • to perform metal(loid)-related phenotyping of the Arabidopsis thaliana metal-sensitive mutant strains[2]
  • as a component of Murashige and Skoog (MS) media to culturing Arabidopsis thaliana seeds[3]
  • to culture Cicer arietinum L. (Chickpea) seeds for Agrobacterium rhizogenes mediated root transformation[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shimpei Uraguchi et al.
Frontiers in plant science, 11, 503-503 (2020-06-06)
For researchers in the plant metal field, the agar reagent used for the solid plate medium is a problematic factor because application of different agar types and even a different lot of the same agar type can mask the plant
Antoine Desrut et al.
Journal of experimental botany, 71(22), 7301-7315 (2020-08-30)
Plants live in close relationships with complex populations of microorganisms, including rhizobacterial species commonly referred to as plant growth-promoting rhizobacteria (PGPR). PGPR are able to improve plant productivity, but the molecular mechanisms involved in this process remain largely unknown. Using
Zhihang Feng et al.
Plant physiology, 184(1), 428-442 (2020-07-01)
Mg2+ is among the most abundant divalent cations in living cells. In plants, investigations on magnesium (Mg) homeostasis are restricted to the functional characterization of Mg2+ transporters. Here, we demonstrate that the splicing factors SUPPRESSORS OF MEC-8 AND UNC-52 1
Malika Affaf Dahmani et al.
Frontiers in plant science, 11, 124-124 (2020-03-17)
Plants live in association with complex populations of microorganisms, including Plant Growth-Promoting Rhizobacteria (PGPR) that confer to plants an improved growth and enhanced stress tolerance. This large and diverse group includes endophytic bacteria that are able to colonize the internal
Extraction and characterization of agar from Australian Pterocladia lucida
Chiovitti A, et al.,
Journal of Applied Phycology, 16(1), 41-48 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service