Skip to Content
Merck
All Photos(4)

Key Documents

A8678

Sigma-Aldrich

Agar

suitable for plant cell culture

Synonym(s):

Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
NACRES:
NA.21

biological source

algae (Rhodophyceae)

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

mp

80-90 °C (1.5% in water)

transition temp

transition temp 27-37 °C (1.5%)

application(s)

agriculture

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Agar or Agar-agar is one of the major components found in the cell wall of red algae. It is a polysaccharide complex, comprising about 70% agarose and 30% agaropectin.

Application

Washed agar has been used:
  • as a supplement (0.9% w/v) to Murashige and Skoog basal medium for plant growth
  • in the preparation of cocultivation medium for Agrobacterium tumefaciens mediated transformation in maize (Zea mays) hybrid Hi-II
  • as a component in the complete medium to regenerate protoplasts of Piriformospora indica
  • as a clean agar medium to culture Synechococcus WH7803 strain

Biochem/physiol Actions

Agar is an ester of sulfuric acid in a linear galactan form, that makes it soluble in hot water and insoluble in cold. Washed agar contain less impurities and low level of nutrients.

Preparation Note

Washed

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alga Zuccaro et al.
Fungal genetics and biology : FG & B, 46(8), 543-550 (2009-04-09)
Piriformospora indica (Basidiomycota, Sebacinales) is a root colonizing fungus which is able to increase biomass and yield of crop plants and to induce local and systemic resistance to fungal diseases and tolerance to abiotic stress. A prerequisite for the elucidation
Andrew D Millard
Methods in molecular biology (Clifton, N.J.), 501, 33-42 (2008-12-11)
Cyanophages are a group of viruses which specifically infect cyanobacteria. The cyanobacteria are predominantly aquatic phototrophic bacteria and the two dominant genera Synechococcus and Prochlorococcus contribute significantly to primary production in the oceans. Cyanophages that infect marine cyanobacteria were first
Lawrence R Griffing
Methods in molecular biology (Clifton, N.J.), 1691, 75-102 (2017-10-19)
In this chapter, approaches to the image analysis of the choreography of the plant endoplasmic reticulum (ER) labeled with fluorescent fusion proteins ("stars," if you wish) are presented. The approaches include the analyses of those parts of the ER that
Juan M Vega et al.
Plant cell reports, 27(2), 297-305 (2007-10-17)
High-frequency transformation of maize (Zea mays L.) using standard binary vectors is advantageous for functional genomics and other genetic engineering studies. Recent advances in Agrobacterium tumefaciens-mediated transformation of maize have made it possible for the public to transform maize using
Queeny Wing-Han Yuen et al.
The open biomedical engineering journal, 5, 39-46 (2011-06-07)
Cervical lymph nodes are common sites of metastatic involvement in head and neck cancers. These lymph nodes are superficially located and palpation is a common practice for assessing nodal hardness and staging cancer which is, however, too subjective and with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service