Merck
Helix 8 is the essential structural motif of mechanosensitive GPCRs.
Nature communications (2019-12-21)
Serap Erdogmus, Ursula Storch, Laura Danner, Jasmin Becker, Michaela Winter, Nicole Ziegler, Angela Wirth, Stefan Offermanns, Carsten Hoffmann, Thomas Gudermann, Michael Mederos Y Schnitzler
ABSTRACT
G-protein coupled receptors (GPCRs) are versatile cellular sensors for chemical stimuli, but also serve as mechanosensors involved in various (patho)physiological settings like vascular regulation, cardiac hypertrophy and preeclampsia. However, the molecular mechanisms underlying mechanically induced GPCR activation have remained elusive. Here we show that mechanosensitive histamine H1 receptors (H1Rs) are endothelial sensors of fluid shear stress and contribute to flow-induced vasodilation. At the molecular level, we observe that H1Rs undergo stimulus-specific patterns of conformational changes suggesting that mechanical forces and agonists induce distinct active receptor conformations. GPCRs lacking C-terminal helix 8 (H8) are not mechanosensitive, and transfer of H8 to non-responsive GPCRs confers, while removal of H8 precludes, mechanosensitivity. Moreover, disrupting H8 structural integrity by amino acid exchanges impairs mechanosensitivity. Altogether, H8 is the essential structural motif endowing GPCRs with mechanosensitivity. These findings provide a mechanistic basis for a better understanding of the roles of mechanosensitive GPCRs in (patho)physiology.
Sigma-Aldrich
Fura 2-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
Luteinizing hormone releasing hormone human acetate salt, ≥98% (HPLC), powder
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Leuprolide acetate salt, ≥98% (HPLC)
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt ≥300,000
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder
Sigma-Aldrich
Levocetirizine dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Desloratadine, powder, ≥98% (HPLC)
Sigma-Aldrich
ML297, ≥98% (HPLC)
Sigma-Aldrich
ATI-2341 trifluoroacetate salt, ≥95% (HPLC)
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Neurotensin, ≥90% (HPLC)
Sigma-Aldrich
ZM 241385, ≥98% (HPLC)