Skip to Content
Merck
All Photos(1)

Key Documents

L0399

Sigma-Aldrich

Leuprolide acetate salt

≥98% (HPLC)

Synonym(s):

des-Gly10-[D-Leu6]-LH-RH ethylamide, Leuprorelin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H84N16O12 · xC2H4O2
CAS Number:
Molecular Weight:
1209.40 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

storage temp.

−20°C

SMILES string

CCNC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@H](CO)NC(=O)[C@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](Cc5c[nH]cn5)NC(=O)[C@@H]6CCC(=O)N6

InChI

1S/C59H84N16O12/c1-6-63-57(86)48-14-10-22-75(48)58(87)41(13-9-21-64-59(60)61)68-51(80)42(23-32(2)3)69-52(81)43(24-33(4)5)70-53(82)44(25-34-15-17-37(77)18-16-34)71-56(85)47(30-76)74-54(83)45(26-35-28-65-39-12-8-7-11-38(35)39)72-55(84)46(27-36-29-62-31-66-36)73-50(79)40-19-20-49(78)67-40/h7-8,11-12,15-18,28-29,31-33,40-48,65,76-77H,6,9-10,13-14,19-27,30H2,1-5H3,(H,62,66)(H,63,86)(H,67,78)(H,68,80)(H,69,81)(H,70,82)(H,71,85)(H,72,84)(H,73,79)(H,74,83)(H4,60,61,64)/t40-,41-,42-,43+,44-,45-,46-,47-,48-/m0/s1

InChI key

GFIJNRVAKGFPGQ-LIJARHBVSA-N

Gene Information

human ... GNRHR(2798)
rat ... Gnrhr(81668)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Leu-Leu-Arg-Pro-NHEt

Application

In mice, leuprolide acetate salt has been used to check the need of estrogen receptor α in spermatogenic cells, for development or function in fertilization.

Biochem/physiol Actions

Glp-His-Trp-Ser-Tyr-Leu-Leu-Arg-Pro-NHEt (des-Gly10-[D-Leu6]-LH-RH) is a GnRH (gonadotropin-releasing hormone) agonist, leuprolide. GnRH (Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2), which is also referred as LHRH (luteinizing hormone-releasing hormone) or gonadorelin, is crucial for mammalian reproduction and is released from hypothalamic neurons. It is responsible for the secretion of gonadotropins, luteinizing hormone (LH) and follicle-stimulating hormone (FSH), from the pituitary glands. Mutations in GnRH are associated with normosmic congenital hypogonadotropic hypogonadism. Low concentration of leuprolide can be used for the treatment of endometriosis, uterine fibroids, central precocious puberty and advanced prostate cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Mahato et al.
Endocrinology, 141(3), 1273-1276 (2000-03-04)
Estrogen receptors alpha (ERalpha) and beta (ERbeta) are ligand-dependent transcription factors and members of the nuclear hormone receptor superfamily encoded by separate genes. Male mice homozygous for a mutation in the gene encoding ERalpha are infertile. To determine whether germ
Michael T Schweizer et al.
Cancer, 119(20), 3610-3618 (2013-08-15)
The current trial evaluated 2 common therapies for patients with advanced prostate cancer, docetaxel and hormonal therapy (HT), in the surgical adjuvant setting. TAX-3501 was a randomized, phase 3, adjuvant study post-radical prostatectomy (RP) in high-risk patients with prostate cancer
Daniela Jakubowicz et al.
Clinical science (London, England : 1979), 125(9), 423-432 (2013-05-22)
In women with PCOS (polycystic ovary syndrome), hyperinsulinaemia stimulates ovarian cytochrome P450c17α activity that, in turn, stimulates ovarian androgen production. Our objective was to compare whether timed caloric intake differentially influences insulin resistance and hyperandrogenism in lean PCOS women. A
Syed Nuruddin et al.
PloS one, 9(8), e103607-e103607 (2014-08-05)
Research on Alzheimer's disease (AD) has indicated an association between hormones of the hypothalamic-pituitary-gonadal (HPG) axis and cognitive senescence, indicating that post meno-/andropausal changes in HPG axis hormones are implicated in the neuropathology of AD. Studies of transgenic mice with
Erin I Lewis et al.
Fertility and sterility, 99(7), 2017-2024 (2013-03-08)
To evaluate novel hormonal therapies in patients with unresectable benign metastasizing leiomyoma (BML) disease. Case series. National Institutes of Health (NIH). Five subjects with the diagnosis of BML based on imaging and/or histopathologic diagnosis. Four patients were treated with single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service