Skip to Content
Merck
All Photos(1)

Key Documents

Z0153

Sigma-Aldrich

ZM 241385

≥98% (HPLC)

Synonym(s):

4-(-2-[7-amino-2-{2-furyl}{1,2,4}triazolo{2,3-a} {1,3,5}triazin-5-yl-amino]ethyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N7O2
CAS Number:
Molecular Weight:
337.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: >15 mg/mL

storage temp.

room temp

SMILES string

Nc1nc(NCCc2ccc(O)cc2)nc3nc(nn13)-c4ccco4

InChI

1S/C16H15N7O2/c17-14-20-15(18-8-7-10-3-5-11(24)6-4-10)21-16-19-13(22-23(14)16)12-2-1-9-25-12/h1-6,9,24H,7-8H2,(H3,17,18,19,20,21,22)

InChI key

PWTBZOIUWZOPFT-UHFFFAOYSA-N

Biochem/physiol Actions

ZM 241385 is a potent selective adenosine A2A antagonist.
ZM 241385 is a potent selective adenosine A2A antagonist. The A2A receptor plays a role in regulating myocardial oxygen consumption and coronary blood flow and is highly expressed in the brain, where it has important roles in the regulation of glutamate and dopamine release. ZM 241385 has neuroprotective effects and is being investigated for use in Parkinson′s and other neurodegenerative disorders.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krystyna Gołembiowska et al.
Neurotoxicity research, 22(2), 150-157 (2012-03-13)
It has been shown that a decreased vesicular monoamine transporter (VMAT2) function and the disruption of dopamine (DA) storage is an early contributor to oxidative damage of dopamine neurons in Parkinson's disease (PD). In our previous study, we demonstrated that
Tarek K Motawi et al.
Molecular and cellular biochemistry, 465(1-2), 89-102 (2019-12-11)
Parkinson's disease (PD) is the second common age-related neurodegenerative disease. It is characterized by control loss of voluntary movements control, resting tremor, postural instability, bradykinesia, and rigidity. The aim of the present work is to evaluate curcumin, niacin, dopaminergic and
Chiara Zoppellaro et al.
PloS one, 8(8), e72648-e72648 (2013-09-10)
Adenosine plays an important role in regulating intestinal motility and inflammatory processes. Previous studies in rodent models have demonstrated that adenosine metabolism and signalling are altered during chronic intestinal inflammatory diseases. However, the involvement of the adenosinergic system in the
Ajith A Welihinda et al.
Cellular signalling, 28(6), 552-560 (2016-02-24)
Inosine is an endogenous purine nucleoside that is produced by catabolism of adenosine. Adenosine has a short half-life (approximately 10s) and is rapidly deaminated to inosine, a stable metabolite with a half-life of approximately 15h. Resembling adenosine, inosine acting through
Krystyna Gołembiowska et al.
Neurotoxicity research, 21(2), 222-230 (2011-08-11)
A(2A) adenosine receptor antagonists have been proposed as a new therapy of PD. Since oxidative stress plays an important role in the pathogenesis of PD, we studied the effect of the selective A(2A) adenosine receptor antagonists 8-(-3-chlorostyryl)caffeine (CSC) and 4-(2-[7-amino-2-(2-furyl)[1,2,4]triazolo[2,3-a][1,3,5]triazin-5-ylamino]ethyl)phenol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service