Skip to Content
Merck
All Photos(1)

Documents

L7795

Sigma-Aldrich

Levocetirizine dihydrochloride

≥98% (HPLC)

Synonym(s):

Levocetirizine dihydrochloride, (-)-Cetirizine dihydrochloride; 2-[2-[4-[(R)-(4-chlorophenyl)-phenyl-methyl]piperazin-1-yl]ethoxy]acetic acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClN2O3·2HCl
CAS Number:
Molecular Weight:
461.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

solubility

H2O: ≥23 mg/mL

storage temp.

room temp

SMILES string

Cl.Cl.OC(=O)COCCN1CCN(CC1)[C@H](c2ccccc2)c3ccc(Cl)cc3

InChI

1S/C21H25ClN2O3.2ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H/t21-;;/m1../s1

InChI key

PGLIUCLTXOYQMV-GHVWMZMZSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Levocetirizine dihydrochloride is the R-enantiomer of cetirizine and is used for treating seasonal allergic rhinitis and chronic idiopathic urticaria. The liver metabolizes 14% of levocetirizine and rest is excreted in urine. The response of levocetirizine to histamine is comparatively higher to cetirizine.

Application

Levocetirizine dihydrochloride has been used as standard in the thin layer chromatography optimization and in preparation of racemic cetirizine standard for subcritical fluid chromatography-tandem mass spectrometry analysis.

Biochem/physiol Actions

Levocetirizine hydrochloride is a nonsedating antihistamine. It is a histamine H1-receptor antagonist, the active isomer of cetirizine. Levocetirizine has high bioavailability, high affinity for and occupancy of the H1 receptor.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigeru Hishinuma et al.
International journal of molecular sciences, 19(12) (2018-12-19)
Cetirizine is a zwitterionic second-generation antihistamine containing R- and S-enantiomers, levocetirizine, and (S)-cetirizine. Levocetirizine is known to have a higher affinity for the histamine H₁ receptors than (S)-cetirizine; ligand-receptor docking simulations have suggested the importance of the formation of a
Dhagla R Choudhary et al.
Scientia pharmaceutica, 80(3), 779-787 (2012-09-26)
A fast-dissolving film containing levocetirizine, a non-sedative antihistamine drug, was developed using pullulan, xanthan gum, propylene glycol, and tween 80 as the base materials. The drug content of the prepared films was within an acceptable limit as prescribed by the
Rapid chiral separation of racemic cetirizine in human plasma using subcritical fluid chromatography-tandem mass spectrometry
Eom HY, et al.
Journal of Pharmaceutical and Biomedical Analysis, 117, 380-389 (2016)
Nurul Atika Razali et al.
BMC complementary and alternative medicine, 18(1), 217-217 (2018-07-18)
Histamine is a well-known mediator involved in skin allergic responses through up-regulation of pro-inflammatory cytokines. Antihistamines remain the mainstay of allergy treatment, but they were found limited in efficacy and associated with several common side effects. Therefore, alternative therapeutic preferences
A double-blind, randomized, single-dose, crossover comparison of levocetirizine with ebastine, fexofenadine, loratadine, mizolastine, and placebo: suppression of histamine-induced wheal-and-flare response during 24 hours in healthy male subjects
Grant JA, et al.
Annals of Allergy, Asthma & Immunology, 88(2), 190-197 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service