Skip to Content
Merck
All Photos(2)

Documents

208752

Sigma-Aldrich

Hexane

≥95%, Laboratory Reagent, suitable for EPA 1613

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21

product name

Hexane, Laboratory Reagent, ≥95%

grade

Laboratory Reagent

Quality Level

Agency

suitable for EPA 1613

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥95%

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

impurities

≤0.005% aromatics (as benzene)
≤0.015% (water)

evapn. residue

≤0.0015%

refractive index

n20/D 1.375 (lit.)

pH

7

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexane is an unbranched alkane that exists in five structural isomeric forms, i.e. hexane, isohexane, 3-methylpentane, neohexane, and 2,3-dimethylbutane. It can be obtained from petroleum by distillation.

Application

Hexane can be used:      
  • As a solvent for the extraction of oil, wax, and fat.     
  • As a non-polar solvent in chromatographic applications for product separation and purification.

It is also used in the preparation of organolithium reagents because it cannot be easily deprotonated.

Other Notes

The article number 208752-4X2.5L will be discontinued. Please order the single bottle 208752-2.5L which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of pre-treatments on the lipid extraction and biodiesel production from municipal WWTP sludge.
Olkiewicz M, et al.
Fuel: The Science and Technology of Fuel and Energy, 141, 250-257 (2015)
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)
Alexandre S Cristino et al.
Nature communications, 5, 5529-5529 (2014-11-21)
Increasing evidence suggests small non-coding RNAs (ncRNAs) such as microRNAs (miRNAs) control levels of mRNA expression during experience-related remodelling of the brain. Here we use an associative olfactory learning paradigm in the honeybee Apis mellifera to examine gene expression changes
Mohammed Anwar et al.
International journal of pharmaceutics, 468(1-2), 158-164 (2014-04-22)
Nano-sized curcumin-loaded super-paramagnetic iron oxide nanoparticles (CUR-OA-SPIONs) were synthesized chemically by co-precipitation method using oleic acid as a stabilizer and Myrj 52 as a surfactant. The synthesized nanoparticles were characterized for their shape, size, surface morphology, electrokinetic potential, magnetic properties
Hans Peter H Arp et al.
Environmental science & technology, 48(19), 11187-11195 (2014-09-13)
Soil quality standards are based on partitioning and toxicity data for laboratory-spiked reference soils, instead of real world, historically contaminated soils, which would be more representative. Here 21 diverse historically contaminated soils from Sweden, Belgium, and France were obtained, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service