Skip to Content
Merck
All Photos(1)

Key Documents

80023449

TSKgel® UP-SW3000 (2 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, diol phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Product Name

TSKgel® UP-SW3000 HPLC Column, phase diol, L × I.D. 15 cm × 4.6 mm, 2 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 0023449

parameter

≤0.5 M salt concentration
0-30% aqueous soluble organic solvents
0.35 mL/min flow rate
10-30 °C temp. range
340 bar pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

2 μm

pore size

250 Å

pH range

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

Application

TSKgel® UP-SW3000 HPLC Column may be used for the separation of:
  • Free payloads and related species from antibody drug conjugates (ADCs) and their aggregates, during their determination by using 1-dimensional (1D) gradient size exclusion chromatography (SEC).
  • Protein biopharmaceuticals in aqueous solutions, during their analysis by SEC.

Other Notes

Related Catagories: Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel Gel Filtration Columns for Proteins and Other Biopolymers, TSKgel Gel Filtration Columns to Analyze Purity and Aggregates, TSKgel Size Exclusion Columns

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extending the limits of size exclusion chromatography: Simultaneous separation of free payloads and related species from antibody drug conjugates and their aggregates.
Goyon A, et al.
Journal of Chromatography A, 1539, 19-29 (2018)
Comprehensive study on the effects of sodium and potassium additives in size exclusion chromatographic separations of protein biopharmaceuticals.
Goyon A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 144, 242-251 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service