Skip to Content
Merck
All Photos(1)

Documents

80023448

TSKgel® UP-SW3000 (2 µm) HPLC Columns

L x I.D. 30 cm × 4.6 mm, diol phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product name

TSKgel® UP-SW3000 HPLC Column, phase diol, L × I.D. 30 cm × 4.6 mm, 2 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 0023448

parameter

≤0.5 M salt concentration
0-30% aqueous soluble organic solvents
0.35 mL/min flow rate
10-30 °C temp. range
340 bar pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 4.6 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

2 μm

pore size

250 Å

pH range

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® UP-SW3000, the new 2 micron UHPLC column with 25 nm pore size, is the latest addition to the renowned TSKgel SW series. TSKgel UP-SW3000 represents the fifth generation of high performance gel filtration columns. They feature the same pore size as the well-established TSKgel G3000SWXL and facilitate method transfer from conventional gel filtration to UHPLC technology. The 30 cm column delivers dramatically increased resolution between fragments, monomers, and aggregates.

Other Notes

Related Catagories: Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel Gel Filtration Columns for Proteins and Other Biopolymers, TSKgel Gel Filtration Columns to Analyze Purity and Aggregates, TSKgel Size Exclusion Columns

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service