Skip to Content
Merck
All Photos(1)

Documents

822854

TSKgel® SuperSW mAb HR (4 µm) HPLC Column

L x I.D. 30 m × 7.8 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® SuperSW mAb HR HPLC Column, phase diol, L × I.D. 30 m × 7.8 mm, 4 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh SuperSW mAb HR, 22854

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1 mL/min flow rate
10-30 °C temperature
120 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 m × 7.8 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

4 μm

pore size

250 Å pore size

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

The TSKgel SuperSW and UltraSW mAb columns consist of three specialized columns designed for the separation and analysis of monoclonal antibodies (mAb). While mAbs can be analyzed using many different modes of HPLC, size exclusion is best to determine the level of aggregation, dimer formation, and fragmentation, making it the best method for heterogeneity studies. TSKgel SuperSW mAb HR columns are optimized for high resolution analysis of mAb monomer and dimer; these 30 cm x 7.8 mm ID slica-based stainless steel columns offer reduced lot-to-lot variation, long column life, reduction of unspecified adsorption, and superior recovery of aggregates. The TSKgel SuperSW mAb columns utilize a unique pore-controlled technology, which produces a shallow calibration curve in the molecular weight region of a typical monoclonal antibody. The calibration curve for the TSKgel SuperSW mAb HR column is similar to that of the industry-leading TSKgel G3000SWxl column and has a shallower slope than the TSKgel UltraSW Aggregate column around the molecular weight range of gamma-globulin. This shallow calibration curve produces high resolution separations.

Application

TSKgel® superSW mAb HR HPLC column may be used in the separation of monoclonal antibodies using size exclusion chromatography (SEC).

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Rau et al.
Molecular cancer therapeutics, 19(7), 1474-1485 (2020-05-21)
The frequent activation of HER3 signaling as a resistance mechanism to EGFR-targeted therapy has motivated the development of combination therapies that block more than one receptor tyrosine kinase. Here, we have developed a novel tetravalent, bispecific single-chain diabody-Fc fusion protein
Alexander Rau et al.
mAbs, 13(1), 1902034-1902034 (2021-03-24)
Dual targeting of surface receptors with bispecific antibodies is attracting increasing interest in cancer therapy. Here, we present a novel bivalent and bispecific antagonistic molecule (Dab-Fc) targeting human epidermal growth factors 2 and 3 (HER2 and HER3) derived from the
null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service