58931
SUPELCOSIL™ LC-18-S (5 µm) HPLC Columns
L × I.D. 15 cm × 4.6 mm, HPLC Column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
eCl@ss:
32110501
Recommended Products
Product Name
SUPELCOSIL™ LC-18-S HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Agency
suitable for USP L1
feature
endcapped
manufacturer/tradename
SUPELCOSIL™
extent of labeling
11.0% carbon loading
parameter
≤70 °C temp. range
400 bar pressure (5801 psi)
technique(s)
HPLC: suitable
L × I.D.
15 cm × 4.6 mm
surface area
170 m2/g
surface coverage
surface coverage 3.1 μmol/m2
matrix
silica gel, spherical particle platform
matrix active group
C18 (octadecyl) phase
particle size
5 μm
pore size
120 Å
application(s)
food and beverages
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
guard cartridge
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
K M Matar et al.
Therapeutic drug monitoring, 21(5), 559-566 (1999-10-16)
A simple, rapid, sensitive, and reproducible high-performance liquid chromatographic (HPLC) method for simultaneous determination of the antiepileptic drugs (ethosuximide, primidone, lamotrigine, phenobarbital, phenytoin, and carbamazepine) and two metabolites (carbamazepine-diol and carbamazepine-epoxide) in human plasma is described. The procedure involves extraction
O Cudina et al.
Farmaco (Societa chimica italiana : 1989), 55(2), 125-127 (2000-04-27)
Fluocortolone and its esters are synthetic corticosteroids used topically in the treatment of various skin disorders. A method that can be successfully used for the separation and determination of fluocortolone pivalate and fluocortolone hexanoate in suppositories was developed. This method
V Stocchi et al.
Analytical biochemistry, 167(1), 181-190 (1987-11-15)
A simple and fast ion pair reversed-phase high-performance liquid chromatographic method has been developed for the simultaneous determination of ATP, ADP, AMP, GTP, GDP, IMP, NADP+, NADPH+, NAD+, NADH, ADP-ribose, inosine, adenosine, hypoxanthine, and xanthine. This method allows us to
G R Granneman et al.
Clinical pharmacokinetics, 29 Suppl 2, 1-8 (1995-01-01)
A rapid and sensitive assay was developed for the measurement of plasma concentrations of zileuton racemate, a potent inhibitor of 5-lipoxygenase. Zileuton and its inactive N-dehydroxylated metabolite were extracted from human, monkey, and rat plasma by use of a solid-phase
K D Dotson et al.
Lipids, 27(11), 933-939 (1992-11-01)
Human milk triacylglycerols were separated by high-performance liquid chromatography. A 5-mu Supelcosil LC-18 column (Supelco, Inc., Bellefonte, PA) was used with acetone/acetonitrile (64:36, vol/vol) as mobile phase. Triacylglycerols were tentatively identified based on theoretical carbon number and relative retention time.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service