Skip to Content
Merck
All Photos(1)

Key Documents

57030-U

Supelco

Visiprep SPE Vacuum Manifold, PTFE Free

standard, 12-port model

Synonym(s):

12-port SPE Vacuum Manifold for cartridge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

material

UHMW PE frit
polypropylene tube

Agency

suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

reg. compliance

suitable for FDA C-010.02

product line

Visiprep

technique(s)

solid phase extraction (SPE): suitable

suitability

suitable for (for PFAS testing)

application(s)

PFAS testing
environmental
food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Visiprep Solid Phase Extraction Vacuum Manifolds andaccessories automate the preparation and cleanup of laboratory liquid samplesby solid phase extraction. Used with a vacuum pump, a Visiprep™ manifold canextract up to 12 or up to 24 samples simultaneously, significantly increasingsample throughput while decreasing sample preparation hands-on time, relativeto liquid-liquid extraction methods. Visiprep vacuum manifolds feature avacuum bleed valve and a stand-alone coverto provide you with precise flow control and safe, easy use.With our flow control valves you can precisely manipulate flow through eachtube by rotating the individual screw-valves built into the cover. Our recentlyoptimized standard manifolds are suitable for SPE methods in the PFAS testingarea. All crucial components that contain PFAS (e.g. PTFE) are substituted byPFAS free alternatives like e.g ultra-high molecular weight Polyethylene (UHMWPE) or stainless steel (needle guides). These ensure low PFAS backgroundcontamination during the SPE process. The glass basin will not dissolve, fog,or discolor when exposed to powerful solvents. The manifold rack accommodatesautosampler or small scintillation vials, 10 mm or 16 mm test tubes, or 1, 2, 5or 10 mL volumetric flasks. An optional plate fits up to ten 20 mLscintillation vials in the 12-port model. The vacuum bleed valve improves control of the vacuum level andensures more secure closure of the valve. This screw-type valve is fitted withan ultra-high molecular weight polyethylene cone shaped seal (now PTFE-free).Tightening the screw ensures a complete seal with no leakage at the valve.Loosening the screw breaks the seal, allowing immediate vacuum release to stopextractions or to lift the manifold cover. Four 2" legs enable you tostand the cover on a work surface with the SPE tubes maintaining their uprightpositions. This prevents solvent guides from resting on the work surface andbecoming damaged or contaminated. Drying attachment on themanifold is sold separately, it installs in minutes and can be used with anyinert gas. The drying attachment also is useful for concentrating recoveredsamples. Gas flow to each tube can be independently adjusted.

Application

Visiprep SPE Vacuum Manifold was used to perform solid phase extraction, to separate polyphenols from grapes, in order to determine the polyphenol content using LC-MS analysis. It was also used for concentration and purification of solvent extracts from surface water and sewage water sediments. Furthermore it was used to extract and determine, estrone in water using solid-phase extraction-enzyme-linked immunosorbent assay (SPE-ELISA).

Features and Benefits

  • Patented screw-type valves within each SPE port for precise flowcontrol
  • Glass basin will not dissolve, fog, or discolor when exposed tosolvents
  • Legs on cover allow user to easily rest cover on work surface whenremoved from the manifold
  • Screw-type solvent-resistant vacuum bleed gauge and valve offerbetter sealing and vacuum control. Valve takes 1/4″ vacuumtubing.
  • PP collection vessel rack accommodates autosampler vials; smallscintillation vials (22.75 mm O.D. recommended); 10 and 16 mm test tubes; and 1, 2, 5, and 10 mL volumetric flasks. An optional plate for 20 mL scintillationvials is available for 12-port models.

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina Mladenov et al.
Journal of cosmetic science, 71(5), 251-262 (2020-10-07)
Caffeine is extensively used in cellulite and hair growth cosmetic products. Regulations in the field of cosmetics require manufacturers to list caffeine in the ingredient list on product labels, but its exact content in these products is not declared. On
Aysha Ferdoushi et al.
Frontiers in oncology, 10, 1601-1601 (2020-09-29)
Schwann cells (SCs), the glial component of peripheral nerves, have been identified as promoters of pancreatic cancer (PC) progression, but the molecular mechanisms are unclear. In the present study, we aimed to identify proteins released by SCs that could stimulate
Nazli Bahrami et al.
The Journal of steroid biochemistry and molecular biology, 200, 105641-105641 (2020-03-11)
The aromatase inhibitors (AIs), letrozole (Femar®/Femara®) and exemestane (Aromasin®), are widely used to treat estrogen receptor (ER) positive breast cancer in postmenopausal patients. In the setting of metastatic breast cancer, these drugs may be used after another causing new responses
Angeles M Alonso Borbalán et al.
Journal of chromatography. A, 1012(1), 31-38 (2003-09-26)
There is considerable interest in the analysis of phenolic compounds due to their involvement in the organoleptic characteristics of foods and drinks, and their contribution to protection against cardiovascular diseases and cancers through their antioxidant activity. This article describes studies
Guangdi Wang et al.
Journal of environmental monitoring : JEM, 14(5), 1353-1364 (2012-03-23)
Endocrine disrupting compounds (EDCs), represented by steroid hormones, organochlorine pesticides (OCPs), polychlorinated biphenyls (PCBs), and bisphenol A have been determined in four sediment cores from the Gulf of Mexico, from New Orleans surface water (Lake Pontchartrain and Mississippi River), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service