Skip to Content
Merck
All Photos(1)

Key Documents

58928-U

Supelco

SUPELCOSIL LC-18-S (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

SUPELCOSIL LC-18-S Reversed-Phase Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L1

product line

SUPELCOSIL

feature

endcapped

manufacturer/tradename

SUPELCOSIL

packaging

1 ea of

extent of labeling

11.0% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

operating pH

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-18-S columns are designed for reliable separations of deoxyribonucleosides and ribonucleosides.

Application


  • Determination of epoxides by high-performance liquid chromatography following derivatization with N,N-diethyldithiocarbamate.: This study discusses the use of SUPELCOSIL LC-18-S HPLC Column for the determination of epoxides in various samples. The research highlights the efficiency of the column in separating epoxide derivatives, showcasing its application in analytical chemistry for accurate and reliable compound analysis. (Dupard-Julien et al., 2007).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mert Ilhan et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 578103-578103 (2014-08-29)
The aim of the present research is to evaluate the antiurolithiatic effect of the various extracts prepared from the fruits of Viburnum opulus L., in regard to its ethnobotanical record. To induce urolithiasis, 70 mg/kg sodium oxalate was injected to the
L Marai et al.
Canadian journal of biochemistry and cell biology = Revue canadienne de biochimie et biologie cellulaire, 61(8), 840-849 (1983-08-01)
Natural triacylglycerols were resolved by high pressure liquid chromatography on Supelcosil LC-18 columns using a gradient of 30-90% propionitrile in acetonitrile as eluting solvent. The effluent was admitted to a quadrupole mass spectrometer via a direct liquid inlet interface. The
Thadikamala Sathish et al.
Pakistan journal of pharmaceutical sciences, 22(1), 68-73 (2009-01-27)
An accurate, simple, reproducible and sensitive RP-HPLC method for the determination of bharangin has been developed and validated. The separation of bharangin and 2-nitroaniline (internal standard) was achieved on Supelcosil LC-18 (3micro, 150 x 4.6 mm i.d.) column using UV
K D Dotson et al.
Lipids, 27(11), 933-939 (1992-11-01)
Human milk triacylglycerols were separated by high-performance liquid chromatography. A 5-mu Supelcosil LC-18 column (Supelco, Inc., Bellefonte, PA) was used with acetone/acetonitrile (64:36, vol/vol) as mobile phase. Triacylglycerols were tentatively identified based on theoretical carbon number and relative retention time.
V Stocchi et al.
Analytical biochemistry, 146(1), 118-124 (1985-04-01)
A simple and rapid method for the determination of ATP, ADP, AMP, NADP+, NAD+, NADPH, and NADH in human erythrocytes is described. A single-step extraction procedure employing alkaline medium and CF 50A Amicon ultrafiltration membranes allows a simultaneous and total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service