Skip to Content
Merck
All Photos(1)

Key Documents

569226-U

Supelco

Discovery® C18 (5 µm) HPLC Columns

L × I.D. 25 cm × 21.2 mm, HPLC Column

Synonym(s):

Discovery RP18 HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

12% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 21.2 mm

surface area

300 m2/g

surface coverage

3 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

180 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Use Discovery® C18 for any method that specifies a C18. The exceptional peak shape, reproducibility, and stability make it the column of choice for all C18 methods from demanding to routine.

Features and Benefits

  • Excellent reproducibility
  • Exceptional peak shape for basic and acidic analytes
  • Stable, low-bleed LC-MS separations
  • Separation of peptides and small proteins
  • Lower hydrophobicity than many comparable C18 columns, providing faster analysis

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Nagano et al.
Journal of dental research, 88(9), 823-828 (2009-09-22)
Mmp-20 and Klk4 are the two key enamel proteases. Can both enzymes process amelogenin to generate the major cleavage products that accumulate during the secretory stage of amelogenesis? We isolated Mmp-20 and Klk4 from developing pig teeth and used them
Yuan-Qing Xia et al.
Rapid communications in mass spectrometry : RCM, 20(12), 1831-1837 (2006-05-18)
A major challenge in selecting an appropriate stationary phase for diastereomeric separation is that it is difficult to predict which of the commercially available stationary phases could achieve the required liquid chromatographic (LC) separation. This work describes the selection and
T R Shantha Kumar et al.
Journal of pharmaceutical and biomedical analysis, 38(1), 173-179 (2005-05-24)
A simple, precise and accurate isocratic reverse-phase liquid chromatography method with programmed wavelength detection has been validated to quantify DRF-4367 and Phenol red, simultaneously for application in rat in situ single pass intestinal perfusion study to assess intestinal permeability of
Yoshinori Masukawa et al.
Journal of lipid research, 49(7), 1466-1476 (2008-03-25)
Ceramides (CERs) in human stratum corneum (SC) play physicochemical roles in determining barrier and water-holding functions of the skin, and specific species might be closely related to the regulation of keratinization, together with other CER-related lipids. Structures of those diverse
Pilar Heredia-Middleton et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 149(4), 552-556 (2008-02-01)
Heat shock protein 70 (HSP70, 70 kDa) is the most commonly expressed protein in response to thermal stress. The extent of its expression is associated with differences in environmental temperatures. We investigated the heat shock response in red blood cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service