Skip to Content
Merck
All Photos(1)

Documents

53914-U

Supelco

Ascentis® Express 90 Å RP-Amide (2.7 μm) HPLC Columns

L × I.D. 15 cm × 2.1 mm, HPLC Column

Synonym(s):

Core-shell (SPP) Fused Core polar embedded HPLC column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Ascentis® Express RP-Amide, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 2.1 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L60

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

parameter

60 °C temp. range
600 bar max. pressure (9000 psi)

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

15 cm × 2.1 mm

surface area

135 m2/g

impurities

<5 ppm metals

matrix

Fused-Core® particle platform
superficially porous particle

matrix active group

amide, alkyl phase

particle size

2.7 μm

pore size

90 Å

operating pH range

2-9

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katia Arena et al.
Journal of separation science, 43(9-10), 1781-1789 (2020-04-03)
Olea europaea, meaning "European olive," is a small tree belonging to the family Oleaceae, occurring in the Mediterranean Basin. Olive oil is an essential component of a balanced diet because of its nutritional value. Among micronutrients, phenolic compounds did show important beneficial effects for human health.
Michael T Furlong et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 926, 36-41 (2013-04-02)
Clopidogrel is prescribed for the treatment of Acute Coronary Syndrome and recent myocardial infarction, recent stroke, or established peripheral arterial disease. A sensitive and reliable high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) assay was developed and validated to enable reliable
Martin Mzik et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1020, 85-89 (2016-04-01)
A simple, rapid and sensitive method based on liquid chromatography-tandem mass spectrometry (LC-MS/MS) has been developed and validated for the quantitative determination in rat plasma of a new candidate for AD treatment, namely PC 48 (a 7-MEOTA-donepezil like compound) in
Erzsébet Oláh et al.
Journal of chromatography. A, 1217(23), 3642-3653 (2010-04-23)
Today sub-2 microm packed columns are very popular to conduct fast chromatographic separations. The mass-transfer resistance depends on the particle size but some practical limits exist not to reach the theoretically expected plate height and mass-transfer resistance. Another approach applies
Maria A Khalikova et al.
Talanta, 130, 433-441 (2014-08-28)
The presented work describes the development of a simple, fast and effective on-line SPE-UHPLC-UV/vis method using fused core particle columns for extraction, separation and quantitative analysis of the nine illegal dyes, most frequently found in chilli-containing spices. The red dyes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service