Skip to Content
Merck
All Photos(1)

Key Documents

1.03725

Supelco

Acetonitrile

≥99.9% (GC), suitable for UHPLC, LiChrosolv®

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
41116004
EC Index Number:
200-835-2
NACRES:
NA.03
technique(s):
HPLC: suitable
LC/MS: suitable
UHPLC: suitable
application(s):
PFAS testing
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

Product Name

Acetonitrile, for UHPLC-MS LiChrosolv®

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 8327
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019

Quality Level

reg. compliance

suitable for FDA C-010.02

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

product line

LiChrosolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

524 °C
973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC: suitable

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

SMILES string

CC#N

application(s)

PFAS testing

format

neat

storage temp.

2-30°C

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrosolv® solvents show high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them suitable for reproducible separations.
Acetonitrile is a polar aprotic solvent, which is used as an intermediate in organic synthesis. It has a low viscosity, high elution strength and miscibility in water that make it viable in many chromatography systems. It can also be utilized in spectrophotometric and fluorimetric technques.

Application

Intended for U/HPLC and LC-MS/MS applications.

Featured as a solvent in screening of benzofurans and ethylphenidate psychostimulants in serum, urine, and hair samples by UHPLC-MS/MS.
The product is an organic solvent, used as a mobile phase/extraction solvent in the ultra-high performance liquid chromatorgraphy - tandem mass spectrometry (UHPLC-MS/MS) analysis of pesticides in tobacco samples.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetonitrile, the polarity chameleon.
P K Zarzycki et al.
Analytical and bioanalytical chemistry, 397(3), 905-908 (2010-04-13)
Intoxication caused by new psychostimulants: analytical methods to disclose acute and chronic use of benzofurans and ethylphenidate
Barcelo B, et al.
International Journal of Legal Medicine, 131(6), 1543-1553 (2017)
Gabrieli Bernardi et al.
Talanta, 161, 40-47 (2016-10-23)
An effective method has been developed and validated for the determination of residues of 55 pesticides in tobacco. The proposed sample preparation method is based on acetonitrile extraction, low-temperature precipitation (LTP) and dispersive solid phase extraction (d-SPE) clean-up. Gas chromatography
Duygu Eryavuz Onmaz et al.
Clinica chimica acta; international journal of clinical chemistry, 497, 120-124 (2019-07-28)
Imatinib has favorable pharmacokinetic properties, but primary and secondary resistance mechanisms may cause a decrease in clinical response over time. There is a positive correlation between serum imatinib concentrations and treatment response. Our aim was to develop a method for
Roberto Nisticò et al.
Nanomaterials (Basel, Switzerland), 9(8) (2019-08-02)
The photodegradation of an aqueous solution of diclofenac (DCF) has been attempted in the presence of hydrogen peroxide and organic/inorganic hybrid magnetic materials under simulated and real solar light. The hybrid magnetic materials have been prepared via coprecipitation synthesis starting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service