Skip to Content
Merck
All Photos(1)

Documents

53500-U

Supelco

Ascentis® Express Guard Cartridge Holder

for use with Ascentis Express Guard Columns, pk of 1

Synonym(s):

HPLC guard column holder for Core-shell (SPP) Fused Core HPLC column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel column

product line

Ascentis®

packaging

pk of 1

technique(s)

HPLC: suitable
LC/MS: suitable

matrix

superficially porous particle

application(s)

food and beverages

compatibility

for use with Ascentis Express Guard Columns

General description

Guard cartridge not included with holder

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service