Skip to Content
Merck
All Photos(1)

Key Documents

51588-U

Supelco

Ascentis® Express 90 Å RP-Amide (2 μm) HPLC Columns

L × I.D. 10 cm × 3 mm UHPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L60

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

parameter

1000 bar max. pressure (14500 psi)
60 °C temp. range

technique(s)

LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

10 cm × 3 mm

surface area

120 m2/g

impurities

<5 ppm (metals)

matrix

Fused-Core particle platform
superficially porous particle

matrix active group

amide, alkyl phase

particle size

2 μm

pore size

90 Å pore size

pH

2-9

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Ascentis® Express 2 μm RP-Amide is a high-speed, high-performance liquid chromatography column based on a new Fused-Core® particle design. The fused core partical provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.4 micron thick porous shell and the small overall particle size of 2-microns. The one-step bonded, end-capped, amide-based polar-embedded stationary phase of Ascentis Express 2 μm RP-Amide provides, a stable, reversed phase packing with decreased hydrophobic character. Ascentis Express 2 μm RP-Amide can be used for basic, acidic, or neutral compounds with orthogonal selectivity from C18 or C8.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Mzik et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1020, 85-89 (2016-04-01)
A simple, rapid and sensitive method based on liquid chromatography-tandem mass spectrometry (LC-MS/MS) has been developed and validated for the quantitative determination in rat plasma of a new candidate for AD treatment, namely PC 48 (a 7-MEOTA-donepezil like compound) in
Erzsébet Oláh et al.
Journal of chromatography. A, 1217(23), 3642-3653 (2010-04-23)
Today sub-2 microm packed columns are very popular to conduct fast chromatographic separations. The mass-transfer resistance depends on the particle size but some practical limits exist not to reach the theoretically expected plate height and mass-transfer resistance. Another approach applies
Hirotaka Kushida et al.
Biomedical chromatography : BMC, 27(12), 1647-1656 (2013-07-03)
Uncaria Hook (UH) alkaloids are involved in the beneficial effects of Yokukansan. However, the pharmacokinetics of UH alkaloids after oral administration of Yokukansan has not yet been sufficiently investigated. Therefore, we developed and validated a sensitive and specific high-performance liquid
Michael T Furlong et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 926, 36-41 (2013-04-02)
Clopidogrel is prescribed for the treatment of Acute Coronary Syndrome and recent myocardial infarction, recent stroke, or established peripheral arterial disease. A sensitive and reliable high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) assay was developed and validated to enable reliable
Fan Sai et al.
Journal of agricultural and food chemistry, 60(8), 1898-1905 (2012-02-15)
A sensitive method has been developed for the simultaneous determination of residues of 25 β₂-agonists and 23 β-blockers in animal foods by high-performance liquid chromatography coupled with linear ion trap mass spectrometry (HPLC-LIT-MS). This method is based on a new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service