Skip to Content
Merck
All Photos(3)

Key Documents

R0395

Sigma-Aldrich

Rapamycin

from Streptomyces hygroscopicus, ≥95% (HPLC), powder, mTOR inhibitor

Synonym(s):

23,27-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine, AY 22989, Sirolimus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H79NO13
CAS Number:
Molecular Weight:
914.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder

Quality Level

Assay

≥95% (HPLC)

form

powder

color

off-white

solubility

ethanol: 2 mM
DMSO: soluble

antibiotic activity spectrum

fungi
yeast

Mode of action

enzyme | inhibits
protein synthesis | interferes

storage temp.

−20°C

SMILES string

CO[C@@H]1C[C@@H](CC[C@H]1O)C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(C)\[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)OC

InChI

1S/C51H79NO13/c1-30-16-12-11-13-17-31(2)42(61-8)28-38-21-19-36(7)51(60,65-38)48(57)49(58)52-23-15-14-18-39(52)50(59)64-43(33(4)26-37-20-22-40(53)44(27-37)62-9)29-41(54)32(3)25-35(6)46(56)47(63-10)45(55)34(5)24-30/h11-13,16-17,25,30,32-34,36-40,42-44,46-47,53,56,60H,14-15,18-24,26-29H2,1-10H3/b13-11+,16-12+,31-17+,35-25+/t30-,32-,33-,34-,36-,37+,38+,39+,40-,42+,43+,44-,46-,47+,51-/m1/s1

InChI key

QFJCIRLUMZQUOT-HPLJOQBZSA-N

Gene Information

human ... FKBP1A(2280)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide
Rapamycin is derived from Streptomyces hygroscopicus . It can inhibit viral replication. This natural anti-fungal antibiotic is a proven anti-aging agent. Rapamycin significantly inhibits mTORC1 regardless of age. It suppresses protein kinase C activity and enhances ion transport in A6 cells. It also suppresses the immune response in membrane and cytosolic preparations. Additionally, it has specific effects on the translation of endogenous mRNAs and inhibits the translation of 5′TOP mRNAs by blocking p70s6k activation in the signaling pathway. Rapamycin exhibits antineoplastic properties.

Application

Rapamycin from Streptomyces hygroscopicus has been used:
  •  to treat BV2 cells or BV2-LC3 cells for autophagy assay
  • as an autophagy activator in the bone marrow mesenchymal stem cells (BM-MSCs) transfected with lentivirus carrier of small interfering RNA for FOXO3 (siFOXO3) to study the effects of Forkhead Box O3 (FOXO3) on the autophagy of BM-MSCs
  • to pre-incubate villi to inhibit the mammalian target of rapamycin complex 1 (mTORC1) signaling

Biochem/physiol Actions

A macrocyclic triene antibiotic that binds to and inhibits the molecular target of rapamycin (mTOR). It forms a complex with FKBP12 that binds to and inhibits the molecular target of rapamycin (mTOR). Rapamycin is a potent immunosuppressant and has anticancer activity.

Features and Benefits

This compound is featured on the PKB/Akt page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapamycin inhibits protein kinase C activity and stimulates Na+ transport in A6 cells
Rokaw MD, et al.
The Journal of Biological Chemistry, 271, 32468-32473 (1996)
FOXO3 is targeted by miR-223-3p and promotes osteogenic differentiation of bone marrow mesenchymal stem cells by enhancing autophagy
Long C, et al.
Human Cell : Official Journal of Human Cell Research Society (2021)
Activation of autophagy pathway suppresses the expression of iNOS, IL6 and cell death of LPS-stimulated microglia cells
Han HE, et al.
Biomolecules & Therapeutics, 21, 21-21 (2013)
Line Heylen et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 60(10), 1505-1511 (2015-02-15)
Transplant recipients are at risk for invasive aspergillosis (IA), associated with a significant mortality rate. Renal transplant-specific risk factors have not been established. Forty-one adult kidney transplant recipients diagnosed with proven or probable IA from 1995 through 2013 were identified
Rapamycin for longevity: opinion article
Blagosklonny, Mikhail V
Aging (Albany. NY.), 11, 8048-8048 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service