Skip to Content
Merck
All Photos(1)

Key Documents

40085

Supelco

Trichloroethylene solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

single component solution

storage temp.

2-8°C

SMILES string

ClC(=CCl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroethylene is a chlorinated aliphatic organic solvent, which can be extensively used as a metal degreaser in industries.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of Zea mays L. in phytoremediation of trichloroethylene
Moccia E, et al.
Environmental Science and Pollution Research International, 24, 11053-11060 (2017)
Hydrodechlorination of trichloroethylene over Pd supported on swellable organically-modified silica (SOMS)
Sohn H, et al.
Applied Catalysis. B, Environmental, 203, 641-653 (2017)
Sung-Wook Jeen et al.
Journal of contaminant hydrology, 144(1), 108-121 (2012-12-19)
Column experiments and numerical simulations were conducted to evaluate the effects of Cr(VI) and dissolved CaCO(3) on the iron reactivity towards trichloroethene (TCE) and Cr(VI) reduction. Column experiments included measurements of iron corrosion potential and characterization of surface film composition
Sara Karami et al.
Occupational and environmental medicine, 70(8), 591-599 (2013-06-01)
The carcinogenic potential of trichloroethylene (TCE) continues to generate much controversy, even after the US Environmental Protection Agency raised its classification to 'carcinogenic to humans'. We conducted a meta-analysis of published cohort and case-control studies exploring occupational TCE exposure in
Sarah J Blossom et al.
Toxicology and applied pharmacology, 269(3), 263-269 (2013-04-10)
Previous studies demonstrated that low-level postnatal and early life exposure to the environmental contaminant, trichloroethylene (TCE), in the drinking water of MRL+/+ mice altered glutathione redox homeostasis and increased biomarkers of oxidative stress indicating a more oxidized state. Plasma metabolites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service