Skip to Content
Merck
All Photos(2)

Key Documents

90935

Supelco

Atrazine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14ClN5
CAS Number:
Molecular Weight:
215.68
Beilstein:
612020
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CCNc1nc(Cl)nc(NC(C)C)n1

InChI

1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)

InChI key

MXWJVTOOROXGIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atrazine is a commonly used herbicide, which belongs to the triazine class of herbicides.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Atrazine maybe used as a reference standard in studies involving the biodegradation of atrazine in polluted soil using different categories of bio-processed materials, which can then be analyzed via high performance liquid chromatography(HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Biodegradation of herbicide (atrazine) in contaminated soil using various bioprocessed materials
Kadian N, et al.
BioTechnology: An Indian Journal, 99, 4642-4647 (2008)
Hermaphroditic, demasculinized frogs after exposure to the herbicide atrazine at low ecologically relevant doses
Hayes.BT, et al.
Proceedings of the National Academy of Sciences of the USA, 99, 5476-5480 (2002)
Ecological risk assessment of atrazine in North American surface waters
Solomon.RK, et al.
Environmental Toxicology and Chemistry / Setac, 15, 31-76 (1996)
Karsten Nödler et al.
Marine pollution bulletin, 70(1-2), 112-118 (2013-03-14)
Despite its EU-wide ban in 2004 the triazine herbicide atrazine (ATR) and its transformation products (TPs) are frequently detected in the aqueous environment. In this study, concentrations of ATR, two of its TPs, and terbuthylazine (TERB) are presented and discussed
Derek W Gammon et al.
Pest management science, 61(4), 331-355 (2005-01-19)
A risk assessment of the triazine herbicide atrazine has been conducted by first analyzing the toxicity database and subsequently estimating exposure. Margins of safety (MOS) were then calculated. Toxicity was assessed in animal studies and exposure was estimated from occupational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service