Skip to Content
Merck
All Photos(1)

Key Documents

48625

Supelco

Vinyl chloride solution

200 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

Agency

ISO 17943

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

single component solution

storage temp.

2-8°C

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here
Vinyl chloride is a colorless gas, which can serve as a monomer for the industrial production of poly(vinyl chloride) bhomopolymer and copolymer. It can also be used in the production of vinyl stearate, vinylidene chloride, tri- and tetra-chloroethylene. It can be obtained via reaction of hydrochloric acid with acetylene derived from calcium carbide.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further supportIntratec Solutions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Luminescent 3d-4f-4d MOF Nanoprobe as a Diagnosis Platform for Human Occupational Exposure to Vinyl Chloride Carcinogen
Hao J-N, et al.
Inorganic Chemistry (2017)
S H Esmaeili Faraj et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 47(14), 2263-2269 (2012-09-01)
A biofiltration process was used for degradation of vinyl chloride as a hazardous material in the air stream. Three biotrickling filters in series-parallel allowing uniform feed and moisture distribution all over the bed were used. Granular activated carbon mixed with
Jun Yan et al.
Applied and environmental microbiology, 78(18), 6630-6636 (2012-07-10)
Dehalococcoides mccartyi strains conserve energy from reductive dechlorination reactions catalyzed by corrinoid-dependent reductive dehalogenase enzyme systems. Dehalococcoides lacks the ability for de novo corrinoid synthesis, and pure cultures require the addition of cyanocobalamin (vitamin B(12)) for growth. In contrast, Geobacter
Kelly Hamonts et al.
Water research, 46(6), 1873-1888 (2012-01-28)
Chlorinated aliphatic hydrocarbons (CAHs) often discharge into rivers as contaminated groundwater baseflow. Biotransformation, sorption and dilution of CAHs in the impacted river sediments have been reported to reduce discharge, but the effect of temporal variations in environmental conditions on the
V S Rukavishnikov et al.
Gigiena i sanitariia, (6)(6), 61-63 (2012-01-19)
The paper provides the results of experimental studies of the impact of vinyl chloride or sublimate intoxication in male albino rats on the functional state of the nervous system of their offspring. Retarded sensorimotor development was revealed in the neonatal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service