Skip to Content
Merck
All Photos(3)

Documents

372145

Sigma-Aldrich

Trichloroethylene

anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%

Synonym(s):

TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

770 °F

contains

40 ppm diisopropylamine as stabilizer

impurities

<0.002% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroethylene (TCE) is a chlorinated ethylene compound commonly used as a dry cleaning and degreasing solvent. Without stabilizers, TCE is unstable and degrades easily in the presence of moisture air, light and heat, it can be minimized by adding diisopropylamine as a stabilizer. Various methods have been reported for its synthesis. One of them is by chlorinating ethylene dichloride. Since TCE is one of the most common industrial solvent it has led it to be a major contaminant found in the ground water. It has adverse effect on human health due to its potential toxicity and carcinogenicity. Its toxicity and carcinogenicity has been studied.
Many methods have been proposed for the mineralization of TCE.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Robert DM and Murphy B.
Chlorinated Solvents: A Forensic Evaluation, 125-131 null
A L Teel et al.
Water research, 35(4), 977-984 (2001-03-10)
Contaminant degradation, stoichiometry, and role of hydroxyl radicals (OH*) in four Fenton's systems were investigated using trichloroethylene (TCE) as a model contaminant. A standard Fenton's system, a modified soluble iron system with a pulse input of hydrogen peroxide, and two
Chlorinated Decreasing Solvents: Physical-Chemical Properties Affecting Aquifer Contamination and Remediation.
Jackson RE and Dwarakanath V.
Ground Water Monitoring & Remediation, 19(4), 102-110 (1999)
E M Waters et al.
Journal of toxicology and environmental health, 2(3), 671-707 (1977-01-01)
Trichloroethylene (TCE) has been an industrial chemical of some importance for the past 50 years. First synthesized by Fischer in 1864, TCE has enjoyed considerable industrial usage as a degreaser and limited medical use as an inhalation anesthetic and analgesic.
Kyung Sun Park et al.
Science advances, 5(8), eaaw7757-eaaw7757 (2019-08-27)
Intrachain charge transport is unique to conjugated polymers distinct from inorganic and small molecular semiconductors and is key to achieving high-performance organic electronics. Polymer backbone planarity and thin film morphology sensitively modulate intrachain charge transport. However, simple, generic nonsynthetic approaches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service