Skip to Content
Merck
All Photos(1)

Key Documents

M6316

Sigma-Aldrich

MRS 1754 hydrate

≥98% (HPLC), solid

Synonym(s):

8-[4-[((4-Cyanophenyl)carbamoylmethyl)oxy]phenyl]-1,3-di(n-propyl)xanthine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N6O4 · xH2O
CAS Number:
Molecular Weight:
486.52 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: soluble >10 mg/mL

storage temp.

room temp

SMILES string

O.CCCN1C(=O)N(CCC)c2[nH]c(nc2C1=O)-c3ccc(OCC(=O)Nc4ccc(cc4)C#N)cc3

InChI

1S/C26H26N6O4.H2O/c1-3-13-31-24-22(25(34)32(14-4-2)26(31)35)29-23(30-24)18-7-11-20(12-8-18)36-16-21(33)28-19-9-5-17(15-27)6-10-19;/h5-12H,3-4,13-14,16H2,1-2H3,(H,28,33)(H,29,30);1H2

InChI key

ZACJDWOPQROWIK-UHFFFAOYSA-N

Application

MRS 1754 hydrate has been used as an adenosine receptor A2B (RA2B) antagonist:
  • to study its effects on endothelial nitric oxide synthase (eNOS) phosphorylation induced by short-term acetate stimulation in human umbilical vein endothelial cells (HUVECs)
  • blastema formation in zebrafish
  • to serve as a positive control in the anti-sickling assay and to study the role of RA2B in the same

Biochem/physiol Actions

MRS 1754 hydrate is a p-cyanoanilide xanthine derivative. It might act as a potential anti-asthmatic drug.
MRS 1754 hydrate is a potent A2B adenosine receptor antagonist.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Manufactured and sold under license from Adenosine Therapeutics LLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[3H] MRS 1754, a selective antagonist radioligand for A2B adenosine receptors
Ji XD, et al.
Biochemical Pharmacology, 61(6), 657-663 (2001)
Vikas Saxena et al.
Cell reports, 39(3), 110727-110727 (2022-04-21)
Regulatory T cell (Treg) lymphatic migration is required for resolving inflammation and prolonging allograft survival. Focusing on Treg interactions with lymphatic endothelial cells (LECs), we dissect mechanisms and functional consequences of Treg transendothelial migration (TEM). Using three genetic mouse models of
X Ji et al.
Biochemical pharmacology, 61(6), 657-663 (2001-03-27)
MRS 1754 [N-(4-cyanophenyl)-2-[4-(2,3,6,7-tetrahydro-2,6-dioxo-1,3-dipropyl-1H-purin-8-yl)-phenoxy]acetamide] is a selective antagonist ligand of A(2B) adenosine receptors. This is the least well-defined adenosine receptor subtype, and A(2B) antagonists have potential as antiasthmatic drugs. For use as a radioligand, MRS 1754, a p-cyanoanilide xanthine derivative, was
Odailson Santos Paz et al.
European journal of medicinal chemistry, 136, 487-496 (2017-05-22)
Adenosine receptors are considered as potential targets for drug development against several diseases. The discovery of subtype 2B adenosine receptors role in erythrocyte sickling process proved its importance to neglected diseases such as sickle cell anemia, which affects approximately 29.000
Vinegar intake enhances flow-mediated vasodilatation via upregulation of endothelial nitric oxide synthase activity
Sakakibara S, et al.
Bioscience, Biotechnology, and Biochemistry, 74(5), 1055-1061 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service