Skip to Content
Merck
All Photos(1)

Key Documents

S6451

Sigma-Aldrich

SCH-442416

≥98% (HPLC)

Synonym(s):

2-(2-Furanyl)-7-[3-(4-methoxyphenyl)propyl]-7H-pyrazolo [4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine; 5-amino-7-(3-(4-methoxyphenyl)propyl)-2-(2 furyl)pyrazolo[4,3-e]-1,2,4-triazolo[1,5-c]pyrimidine; 5-Amino-7-[3-(4-methoxy)phenylpropyl]-2-(2-furyl)-pyrazolo[4,3-e]-1,2,4-triazolo[1,5-c]pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N7O2
CAS Number:
Molecular Weight:
389.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

light tan to tan

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(CCCn2ncc3c2nc(N)n4nc(nc34)-c5ccco5)cc1

InChI

1S/C20H19N7O2/c1-28-14-8-6-13(7-9-14)4-2-10-26-18-15(12-22-26)19-23-17(16-5-3-11-29-16)25-27(19)20(21)24-18/h3,5-9,11-12H,2,4,10H2,1H3,(H2,21,24)

InChI key

AEULVFLPCJOBCE-UHFFFAOYSA-N

Application

SCH-442416 has been used:
  • as a selective adenosine A2A antagonist to study its effects on cyclic adenosine monophosphate (cAMP) production in T/C-28a2 chondrocytes or hFOB 1.19 osteoblasts 
  • as a presynaptic A2A receptor antagonists to study its effects on receptor blockade on δ9-Tetrahydrocannabinol (THC) self-administration in squirrel monkeys
  • as a presynaptic A2A receptor antagonists to study its effects on cocaine-induced locomotion and cocaine-seeking in mice

Biochem/physiol Actions

SCH-442416 potent and selective Adenosine A2a antagonist. SCH-442416 is >10,000-fold selective for A2a compared to other adensosine receptors.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrizio Vincenzi et al.
PloS one, 8(5), e65561-e65561 (2013-06-07)
Adenosine receptors (ARs) have an important role in the regulation of inflammation and their activation is involved in the inhibition of pro-inflammatory cytokine release. The effects of pulsed electromagnetic fields (PEMFs) on inflammation have been reported and we have demonstrated
Mallikarjunarao Ganesana et al.
PloS one, 13(5), e0196932-e0196932 (2018-05-26)
Adenosine is an important neuromodulator in the central nervous system, and tissue adenosine levels increase during ischemic events, attenuating excitotoxic neuronal injury. Recently, our lab developed an electrochemical fast-scan cyclic voltammetry (FSCV) method that identified rapid, spontaneous changes in adenosine
Ying Wang et al.
ACS chemical neuroscience, 10(4), 1941-1949 (2018-09-27)
Adenosine is an endogenous neuroprotectant that modulates vasodilation in the central nervous system. Oxygen changes occur when there is an increase in local cerebral blood flow and thus are a measure of vasodilation. Transient oxygen events following rapid adenosine events
Effects of adenosine A 2A receptor antagonists on cocaine-induced locomotion and cocaine seeking
Haynes N S, et al.
Psychopharmacology, 236(2), 699-708 (2019)
Xiaohan Xu et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 66(4), 353-364 (2017-01-12)
Neutrophil chemotaxis plays an essential role in recruiting neutrophils to sites of inflammation. Neutrophil chemotaxis is suppressed both after exposure to lipopolysaccharide (LPS) in vitro and during clinical and experimental endotoxemia, leading to serious consequences. Adenosine (ADO) is a potent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service