Skip to Content
Merck
All Photos(1)

Key Documents

M228

Sigma-Aldrich

MRS 1220

solid

Synonym(s):

9-Chloro-2-(2-furanyl)-5-((phenylacetyl)amino)-[1,2,4]triazolo[1,5-c]quinazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14ClN5O2
CAS Number:
Molecular Weight:
403.82
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

DMSO: 2 mg/mL
2-hydroxypropyl-β-cyclodextrin: insoluble
H2O: insoluble
ethanol: insoluble

SMILES string

Clc1ccc2nc(NC(=O)Cc3ccccc3)n4nc(nc4c2c1)-c5ccco5

InChI

1S/C21H14ClN5O2/c22-14-8-9-16-15(12-14)20-25-19(17-7-4-10-29-17)26-27(20)21(23-16)24-18(28)11-13-5-2-1-3-6-13/h1-10,12H,11H2,(H,23,24,28)

InChI key

TWWFAXQOKNBUCR-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

MRS1220 is a putative A3 adenosine receptor antagonist. MRS 1220 was found to be competitive in saturation binding studies using the agonist radioligand 125I AB-MECA at cloned human brain A3 receptors expressed in HEK-293 cells. Antagonism was demonstrated in functional assays consisting of agonist-induced inhibition of adenylate cyclase and the stimulation of binding of 35S guanosine 5′-O-(3-thiotriphosphate (35S GTP-gamma-S) to the associated G-proteins. MRS 1220 and MRS 1191, with KB values of 1.7 and 92 nM, respectively, proved to be highly selective for human A3 receptor vs human A1 receptor-mediated effects on adenylate cyclase. In addition, MRS 1220 reversed the effect of A3 agonist-elicited inhibition of tumor necrosis factor-alpha formation in the human macrophage U-937 cell line, with an IC50 value of 0.3 μM.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naunyn-Schmiedeberg'S Archives of Pharmacology, 364, 225-234 (2000)
Pnina Fishman et al.
Arthritis research & therapy, 8(1), R33-R33 (2006-03-02)
The anti-inflammatory effect of adenosine was previously found to be mediated via activation of the A3 adenosine receptor (A3AR). The aim of the present study was to decipher the molecular mechanism involved with the inhibitory effect of IB-MECA, an A3AR
Y C Kim et al.
Journal of medicinal chemistry, 39(21), 4142-4148 (1996-10-11)
The adenosine antagonist 9-chloro-2-(2-furanyl)[1,2,4]triazolo[1,5-c]quinazolin-5-amine (CGS15943) binds to human A3 receptors with high affinity (Ki = 14 nM), while it lacks affinity at rat A3 receptors. Acylated derivatives of the 5-amino group and other modifications were prepared in an effort to
K A Jacobson et al.
Neuropharmacology, 36(9), 1157-1165 (1997-11-19)
The effects of putative A3 adenosine receptor antagonists of three diverse chemical classes (the flavonoid MRS 1067, the 6-phenyl-1,4-dihydropyridines MRS 1097 and MRS 1191, and the triazoloquinazoline MRS 1220) were characterized in receptor binding and functional assays. MRS1067, MRS 1191
Seyyed Mehdi Jafari et al.
Journal of cellular biochemistry, 118(9), 2909-2920 (2017-02-24)
Numerous studies have demonstrated the role of A3 adenosine receptor (A3AR) and signaling pathways in the multiple aspects of the tumor. However, there is a little study about the function of A3AR in the biological processes of cancer stem cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service