Skip to Content
Merck
All Photos(4)

Key Documents

XNAS2

Sigma-Aldrich

Extract-N-Amp Seed PCR Kit

sufficient for 100 extractions, sufficient for 100 amplifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 100 amplifications
sufficient for 100 extractions
sufficient for 100 reactions

Quality Level

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

application(s)

agriculture

shipped in

dry ice

storage temp.

−20°C

General description

The Extract-N-Amp Seed PCR Kit provides all reagents necessary to rapidly extract DNA from a wide variety of plant seeds and amplify targets of interest by PCR. A novel extraction method eliminates the need for long enzymatic digestions. The kit also includes a specially formulated hot start PCR reaction mix for amplification directly from the extract.

The kit comes with validated protocols to extract and amplify genomic DNA from corn, wheat, sunflower, cotton, soybean, sorghum, canola, and Arabidopsis seeds. Additional protocols are available. In a typical procedure, genomic DNA is extracted from a sample that has been ground and incubated in the Seed Preparation Solution and Extraction Solution for 10 minutes at room temperature. The sample is heated to 95 °C for 3 minutes and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and the Extract-N-Amp PCR ReadyMix, included in the kit.

Application

Extract-N-Amp seed PCR Kit has been used for DNA extraction. It has aslo been used for extraction and amplification of DNA from symptomatic wheat seeds.

Features and Benefits

  • Perfect for quick genotyping results
  • Rapid extraction of genomic DNA for PCR in 15 minutes
  • No long enzymatic digestions
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • Compatible with a wide variety of seed sources
  • Extracts are stable at 4 °C for atleast 6 months, allowing for multiple reassays
  • Phenol/chloroform extraction or alcohol precipitation not required
  • Column purification or centrifugation not required

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brigitte Marazzi et al.
American journal of botany, 93(2), 288-303 (2006-02-01)
Senna (Leguminosae) is a large, widespread genus that includes species with enantiostylous, asymmetric flowers and species with extrafloral nectaries. Clarification of phylogenetic relationships within Senna based on parsimony analyses of three chloroplast regions (rpS16, rpL16, and matK) provides new insights
Helen L Rose et al.
PloS one, 6(7), e22668-e22668 (2011-08-06)
Eight DNA extraction products or methods (Applied Biosystems PrepFiler Forensic DNA Extraction Kit; Bio-Rad Instagene Only, Bio-Rad Instagene & Spin Column Purification; EpiCentre MasterPure DNA & RNA Kit; FujiFilm QuickGene Mini80; Idaho Technologies 1-2-3 Q-Flow Kit; MoBio UltraClean Microbial DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service