Skip to Content
Merck
All Photos(1)

Key Documents

XNAR

Sigma-Aldrich

Extract-N-Amp Plant PCR Kit

sufficient for 1000 extractions, sufficient for 1000 amplifications

Synonym(s):

Plant direct PCR kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 1000 amplifications
sufficient for 1000 extractions

Quality Level

feature

hotstart

technique(s)

PCR: suitable

color

colorless

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Extract-N-Amp Plant PCR Kits are intended for direct PCR (polymerase chain reaction). This kit contains all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. It is a novel Extraction Solution that eliminates the need for conventional freezing of plant tissues.

Application

Extract-N-Amp Plant PCR Kit has been used for DNA extraction from fresh mushrooms.
Extract-N-Amp Plant PCR Kit is suitable for use in genotyping.

It has been used forDNA extraction from fresh mushrooms.

Features and Benefits

  • Plant genomic DNA can be extracted for PCR in under 15 minutes using a method.
  • No freezing, mechanical disruption, organic extraction, column purification or precipitation is necessary
  • A specially formulated PCR ReadyMix is provided for use with the extracted DNA
  • The Hot Start antibody ensures highly specific PCR amplification of genomic DNA
  • Suitable for high-throughput plant genetic analysis.
  • Lengthy enzymatic digestions are not required, saving time and effort.
  • The extracted DNA remains stable at 4 °C for atleast 6 months.

Components

Extract-N-Amp Plant PCR Kits contain Dilution solution, Extraction solution, and Extract-N-Amp PCR Reaction Mix (2X) (buffer, salts, dNTPs, Taq polymerase, and JumpStart Taq antibody)

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.
The Extract-N-Amp Plant PCR Kits are for laboratory use only. Not for drug, household, or other uses.

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison Heffer et al.
EvoDevo, 2(1), 7-7 (2011-03-03)
Tremendous progress has been made in the field of evo-devo through comparisons of related genes from diverse taxa. While the vast number of species in nature precludes a complete analysis of the molecular evolution of even one single gene family
M Riahi et al.
Genetics and molecular research : GMR, 9(3), 1334-1342 (2010-07-21)
Three DNA extraction protocols were compared for their ability to yield DNA from the leaves of herbarium specimens of nine species from nine genera of the Papilionoideae. We tested two protocols that use classic procedures for lysis and purification with
Vasilios M E Andriotis et al.
Plant physiology, 160(3), 1175-1186 (2012-09-04)
Mature seeds of both the high-starch starch-excess1 (sex1) mutant and the almost starchless phosphoglucomutase1 mutant of Arabidopsis (Arabidopsis thaliana) have 30% to 40% less lipid than seeds of wild-type plants. We show that this is a maternal effect and is
Jian-Feng Li et al.
Plant methods, 6(1), 1-1 (2010-02-26)
Research in plant molecular biology involves DNA purification on a daily basis. Although different commercial kits enable convenient extraction of high-quality DNA from E. coli cells, PCR and agarose gel samples as well as plant tissues, each kit is designed
Matthew E Smith et al.
PloS one, 8(1), e55160-e55160 (2013-02-06)
Ectomycorrhizal (ECM) plants and fungi can be diverse and abundant in certain tropical ecosystems. For example, the primarily paleotropical ECM plant family Dipterocarpaceae is one of the most speciose and ecologically important tree families in Southeast Asia. Pakaraimaea dipterocarpacea is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service