Skip to Content
Merck
All Photos(1)

Key Documents

D5688

Sigma-Aldrich

Dilution Solution

Extract-N-Amp Plant Dilution Solution

Synonym(s):

Direct PCR, Direct PCR dilution solution for plants, Extract‑N‑Amp Dilution Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.55

Quality Level

form

liquid

storage temp.

−20°C

General description

Dilution Solution is intended for use with Sigma′s Extract-N-Amp Plant PCR Kit. Once the DNA is extracted from the leaf tissue, the Dilution Solution is added to the extract to neutralize inhibitory substances. The extract is then ready for PCR.

Application

Dilution Solution (Extract-N-Amp Plant Dilution Solution) has been used in direct PCR amplification and to prepare template DNA.

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose L Ramirez et al.
Journal of insect science (Online), 8, 1-7 (2008-04-01)
Homalodisca vitripennis (Germar) (Hemiptera: Cicadellidae), the glassy-winged sharpshooter, is one of the most important vectors of the bacterium, Xylella fastidiosa subsp. piercei (Xanthomonadales: Xanthomonadaceae) that causes Pierce's Disease in grapevines in California. In the present study we report a new
Molecular Diagnosis of Urinary Tract Infections by Semi-Quantitative Detection of Uropathogens in a Routine Clinical Hospital Setting.
van der Zee A
PLoS ONE, 11(3), e0150755-e0150755 (2016)
Anneke van der Zee et al.
BMC infectious diseases, 14, 27-27 (2014-01-16)
Resistance to carbapenem antibiotics is emerging worldwide among Enterobacteriaceae. To prevent hospital transmission due to unnoticed carriage of carbapenemase producing micro-organisms in newly admitted patients, or follow-up of patients in an outbreak setting, a molecular screening method was developed for
Daniela Hegebarth et al.
Plant, cell & environment, 40(9), 1761-1776 (2017-05-10)
The aliphatic waxes sealing plant surfaces against environmental stress are generated by fatty acid elongase complexes, each containing a β-ketoacyl-CoA synthase (KCS) enzyme that catalyses a crucial condensation forming a new C─C bond to extend the carbon backbone. The relatively
Anneke van der Zee et al.
BMC research notes, 4, 150-150 (2011-05-28)
To facilitate automation, a novel DNA extraction method for MRSA was adopted. The MRSA specific chromosome-SCCmec PCR was adapted, additional primers were added, and the performance was validated. From various laboratories in The Netherlands we received a total of 86

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service