Skip to Content
Merck

Skip To

WTA2

Complete Whole Transcriptome Amplification Kit

DNA polymerase included, Complete Kit with optimized enzyme to amplify total RNA in <4 hours, no 3′ bias

Synonym(s):

transcriptome amplification kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.55
UNSPSC Code:
41121800
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

technique(s)

whole genome amplification: suitable

dilution

(WTA)

input

purified RNA

shipped in

wet ice

storage temp.

−20°C

General description

WTA2 is optimized to amplify RNA from formalin-fixed, paraffin-embedded (FFPE) and other damaged or degraded samples. Whole Transcriptome Amplification (WTA) technology, allows for representative amplification of low nanogram quantities of total RNA in less than 4 hours without 3′-bias. Amplification products are suitable for applications such as qPCR, micro array analysis, and cloning. The WTA2 kit contains the polymerase needed to amplify the cDNA library.

Application

Suitable for use with downstream applications including:
  • qPCR
  • microarray analysis
  • cloning
Complete Whole Transcriptome Amplification Kit is used for the following applications:
  • To establish a protocol for the simultaneous analysis of DNA and RNA viruses present in pig feces using process controlled deep sequencing.[1]
  • Reverse transcription and cDNA amplification[2][3][4]
  • For the synthesis and amplification of cDNA library using Genomic RNA released from immunocaptured PPV particles[5]
  • Nucleic Acid Preparation and Deep Sequencing (The extracted nucleic acids were randomly primed for cDNA synthesis)[6]
  • Amplification of RNA andssDNA viruses for Viral metagenomics from low-input clinical samples

Biochem/physiol Actions

The WTA2 process involves two steps. In the first step, sample RNA is reverse transcribed with non-self-complementary primers composed of a quasi-random 3′ end and a universal 5′ end. During this process, displaced single strands serve as new templates for primer annealing and extension. The resultant cDNA library, comprised of random, overlapping 100 - 1000 base fragments flanked by universal end sequence. The 2nd step amplifies the cDNA library by PCR using WTA2 polymerase and a universal end primer to produce WTA2 product.

Features and Benefits

  • Achieve up to 10,000x amplification in less than 4 hours with less than 30 minutes of "hands on" time required
  • Only 20 pg of total RNA template is required to amplify suitable cDNA for microarray profiling
  • Contains all needed components for cDNA amplification
  • Achieve linear amplification of expressed genes and exons without 3′ or 5′ bias
  • Effectively amplifies single cell or low input RNA, including mRNA and total RNA from any animal, plant, or microorganism

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WGA2SEQRWTA1
technique(s)

whole genome amplification: suitable

technique(s)

whole genome amplification: suitable

technique(s)

whole genome amplification: suitable

technique(s)

whole genome amplification: suitable

input

purified RNA

input

purified DNA

input

purified RNA

input

purified DNA

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

dilution

(WTA)

dilution

(WGA)

dilution

(WTA)

dilution

(WTA)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Library Synthesis Enzyme
    SDS

  • Library Synthesis Solution
    SDS

  • Amplification Mix
    SDS

  • Library Synthesis Buffer
    SDS

  • Water, Nuclease-Free Water, for Molecular Biology
    SDS

  • Amplification Enzyme
    SDS

  • Deoxynucleotide Mix, 10 mM, Molecular Biology Reagent
    SDS

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What are the differences between WTA1 and WTA2?

    1 answer
    1. WTA2 is designed to amplify RNA from formalin-fixed, paraffin-embedded (FFPE) and other damaged or degraded samples. Here are the main differences between the two kits:

      1. WTA1 was developed by Rubicon. WTA2 was developed by Sigma-Aldrich.
      2. The library synthesis primers for WTA2 are different and can prime more frequently, which is crucial for degraded RNAs.
      3. The library synthesis conditions are different due to the change in the primers.
      4. There are improved cycling conditions in WTA2 to optimize amplification of both high AT templates and high GC templates.
      5. WTA1 does not contain an amplification enzyme, which needs to be supplied separately. WTA2 includes the amplification enzyme.

      Helpful?

Reviews

No rating value

Active Filters