SEQR
SeqPlex RNA Amplification Kit
For use with high throughput sequencing technologies
Synonym(s):
WGA kit, Whole transcriptome amplification
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
41121800
NACRES:
NA.55
Recommended Products
technique(s)
whole genome amplification: suitable
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
The SeqPlex™ RNA Amplification Kit for whole transcriptome amplification (WTA) is designed to facilitate next-generation sequencing (NGS) from small quantities or from degraded/highly fragmented RNA (e.g. RNA from formalin-fixed paraffin-embedded (FFPE) tissue samples). The SeqPlex kit allows the user to pre-amplify these and other small quantity/highly fragmented RNA samples for input into a NGS workflow. It also facilitates the amplification of non-polyA tailed RNA isolated from tissue, cultured cells, formalin-fixed samples, or serum while maintaining patterns of differential expression found in the unamplified sample.
Application
SeqPlex™ RNA Amplification Kit has been used to amplify poly(A)-selected RNA.
Features and Benefits
- Low quantities of total RNA random priming technology amplifies fragmented or intact RNA from all sources including FFPE and RIP.
- Semi-degenerate library primer design for more complete transcriptome coverage and efficient priming.
- No need to fragment DNA before sequencing.
- Amplifies ds-cDNA in 8 hours or less.
- Compatible with all next generation sequencing platforms except Pacific Bioscience.
Other Notes
SEQR-500RXN is manufactured on-demand. Contact us for more information.
Legal Information
SeqPlex is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Grégory Seumois et al.
Nature immunology, 15(8), 777-788 (2014-07-07)
A characteristic feature of asthma is the aberrant accumulation, differentiation or function of memory CD4(+) T cells that produce type 2 cytokines (TH2 cells). By mapping genome-wide histone modification profiles for subsets of T cells isolated from peripheral blood of
Giuliana P Mognol et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(13), E2776-E2785 (2017-03-12)
T-cell exhaustion is a progressive loss of effector function and memory potential due to persistent antigen exposure, which occurs in chronic viral infections and cancer. Here we investigate the relation between gene expression and chromatin accessibility in CD8+ tumor-infiltrating lymphocytes
Kathryn D Tuttle et al.
Nature communications, 9(1), 2650-2650 (2018-07-10)
During development in the thymus, invariant natural killer T (iNKT) cells commit to one of three major functionally different subsets, iNKT1, iNKT2, and iNKT17. Here, we show that T cell antigen receptor (TCR) signal strength governs the development of iNKT
DNA Sequencing Research Group (DSRG): Evaluation of RNA Amplification Kits at Subnanogram Input Amounts of Total RNA for RNA-Seq
Nicolet C, et al.
Journal of biomolecular techniques : JBT, 24, S70-S70 (2013)
Isaac Engel et al.
Nature immunology, 17(6), 728-739 (2016-04-19)
Natural killer T cells (NKT cells) have stimulatory or inhibitory effects on the immune response that can be attributed in part to the existence of functional subsets of NKT cells. These subsets have been characterized only on the basis of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service