Skip to Content
Merck
All Photos(8)

Key Documents

WH0002597M1

Sigma-Aldrich

Monoclonal Anti-GAPDH antibody produced in mouse

clone 3C2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-G3PD, Anti-GAPD, Anti-MGC88685, Anti-glyceraldehyde-3-phosphate dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)

General description

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH), a classic glycolytic enzyme, is a multi-functional protein. It is mainly present in the cytoplasm. GAPDH is ubiquitously expressed. The GAPDH gene is located on human chromosome 12.
The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome. (provided by RefSeq)

Immunogen

GAPDH (NP_002037, 226 a.a. ~ 335 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GKLTGMAFRVPTANVSVVDLTCRLEKPAKYDDIKKVVKQASEGPLKGILGYTEHQVVSSDFNSDTHSSTFDAGAGIALNDHFVKLISWYDNEFGYSNRVVDLMAHMASKE

Application

Monoclonal Anti-GAPDH antibody produced in mouse has been used in western blot, (1:5000), and (1:2,000).

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) participates in energy metabolism. It might have extra glycolytic roles in DNA repair. GAPDH plays a key role in glycolysis and several non-glycolytic actions. GAPDH binds to microtubules and regulates microtubule bundling and aids in membrane fusion. GAPDH participates in gene transcription, DNA replication, and nuclear RNA export. GAPDH acts as a uracil DNA glycosylase (UDG) that plays a role in DNA repair. It also acts as a mediator for cell death. GAPDH may play a role in Alzheimer′s disease (AD). It can be a potential therapeutic target in chemotherapy. Overexpression of the GAPDH gene in the T cell lineage is associated with angioimmunoblastic T cell lymphoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Zhan et al.
Experimental and therapeutic medicine, 13(5), 2473-2479 (2017-06-02)
Kidney cancer is among the most important causes of cancer-associated mortality worldwide. The present study aimed to evaluate protein kinase C α (PKCα) expression in kidney cancer tissues and cell lines, and its significance in apoptosis and migration. Expression of
Xu Han et al.
Journal of cellular biochemistry, 120(8), 12966-12976 (2019-04-20)
Endocrine therapy resistance represents a major challenge to the successful treatment of patients with breast cancer. The development of tamoxifen resistance commonly occurrs during the treatment of patients with breast cancer whereas its underlying mechanisms remain elusive. Here, we found
Le Lu et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 68(1), 13-19 (2013-11-12)
Abnormal microRNA expression is a common and important feature of human malignancies. Matrix metalloproteinase 2 (MMP2), which has been reported in several cancers, plays important roles in cancer progression. However, the microRNA regulatory mechanism on MMP2 expression remains unclear. In
Makoto R Hara et al.
Biochimica et biophysica acta, 1762(5), 502-509 (2006-04-01)
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a classic glycolytic enzyme, and accumulating evidence has suggested that GAPDH is a multi-functional protein. In particular, its role as a mediator for cell death has been highlighted. For the last decade, many groups reported that
Nan Liu et al.
OncoTargets and therapy, 12, 6393-6406 (2019-09-10)
FAM163A, also called neuroblastoma-derived secretory protein (NDSP) or C1ORF76, was newly found on chromosome 1q25.2. Previous studies of FAM163A focused on its expression and function in neuroblastoma. However, using an online database, we found that FAM163A may predict poor prognosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service