Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300645

Sigma-Aldrich

Anti-GAPDH Antibody

rabbit polyclonal

Synonym(s):

Anti-G3PD, Anti-GAPD, Anti-MGC88685

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-GAPDH antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~37 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(P-A-K-Y-D)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)

General description

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a multifunctional protein , that is located on chromosome 12p13. It is basically called as a glycolytic enzyme.

The antibody detects endogenous level of total GAPDH protein.

Immunogen

Peptide sequence around aa. 252-256 (P-A-K-Y-D), according to the protein NP_002037.2

Application

Anti-GAPDH antibody has been used in western blotting.{60,61}

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) plays an important role in DNA repair. It is essential for the sixth step of glycolysis. It participates in transcriptional and posttranscriptional gene regulation. It also controls intracellular membrane trafficking and cell death. Accumulation of GAPDH results in mitochondrial dysfunction.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Has both glyceraldehyde-3-phosphate dehydrogenase and nitrosylase activities, thereby playing a role in glycolysis and nuclear functions, respectively. Participates in nuclear events including transcription, RNA transport, DNA replication and apoptosis. Nuclear functions are probably due to the nitrosylase activity that mediates cysteine S-nitrosylation of nuclear target proteins such as SIRT1, HDAC2 and PRKDC By similarity. Glyceraldehyde-3-phosphate dehydrogenase is a key enzyme in glycolysis that catalyzes the first step of the pathway by converting D-glyceraldehyde 3-phosphate (G3P) into 3-phospho-D-glyceroyl phosphate.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Túlio de Almeida Hermes et al.
International journal of experimental pathology, 105(2), 75-85 (2024-03-13)
Duchenne muscular dystrophy (DMD) occurs due to genetic mutations that lead to a deficiency in dystrophin production and consequent progressive degeneration of skeletal muscle fibres, through oxidative stress and an exacerbated inflammatory process. The flavonoid trilobatin (TLB) demonstrates antioxidant and
Yanwu Hu et al.
Molecular medicine reports, 13(3), 2899-2903 (2016-01-29)
Icariin, a flavonoid isolated from the traditional Chinese herbal medicine Epimedium brevicornum Maxim, has been shown to possess anti-inflammatory, anti‑oxidant and anti-atherosclerotic activities in vivo and in vitro. The aim of the present study was to investigate the effects of
Qian Zhang et al.
Nephron, 146(6), 624-636 (2022-05-09)
The change of podocyte morphology is a pathologic feature of chronic kidney disease. Several studies have suggested that vitamin D plays a role in the protection of podocytes, but the underlying mechanism remains unclear. The effects of paricalcitol on podocyte
Lin Zhu et al.
Neuroimmunomodulation, 29(4), 439-449 (2022-05-25)
Neuroinflammation subsequent to traumatic brain injury (TBI) is important for the recovery of patients and is associated with neurodegenerative changes post-TBI. The tripartite motif containing 44 (TRIM44) protein is an E3 ligase involved in the regulation of immune function with
Glyceraldehyde-3-phosphate Dehydrogenase (GAPDH) Aggregation Causes Mitochondrial Dysfunction during Oxidative Stress-induced Cell Death
Nakajima H, et al.
The Journal of Biological Chemistry (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service