Skip to Content
Merck
All Photos(3)

Key Documents

AB2302

Sigma-Aldrich

Anti-GAPDH Antibody

from chicken, purified by affinity chromatography

Synonym(s):

glyceraldehyde 3-phosphate dehydrogenase, aging-associated gene 9 protein, glyceraldehyde-3-phosphate dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, horse, rat

species reactivity (predicted by homology)

canine (based on 100% sequence homology), equine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient
wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)
mouse ... Gapdh(14433)

General description

Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) is an ubiquitous glycolytic enzyme present in reasonably high levels in almost all tissues. As a ′house-keeping′ enzyme, it catalyzes the synthesis of 1,3-biphosphoglycerate, a "high energy" intermediate used for the synthesis of ATP. Besides its cytoplasmic action in metabolism it is also involved in the initial stages of apoptosis or oxidative stress response where GAPDH is translocated to the nucleus. Such actions may reflect the role of GAPDH in DNA repair or as one nuclear carrier for apoptotic molecules. GAPDH has also been found to bind specifically to proteins implicated in the pathogenesis of a variety of neurodegenerative disorders including the beta-amyloid precursor protein and the huntingtin protein where decreased function of GAPDH is associated with Alzheimer′s and Huntington′s disease fibroblasts. GAPDH has also been identified as a potential target for nitric oxide (NO)-mediated cellular toxicity. The complete and functional enzyme is a tetramer with each of four identical subunits occupying the vertex of a tetrahedron. Binding domains also include one principally interacting with NAD+ and another interacting with glyceraldehyde 3-phosphate (GAP).

Specificity

This antibody recognizes GAPDH.

Immunogen

Epitope: WARS interacting region
KLH-conjugated linear peptide corresponding to the WARS interacting region of human GAPDH.

Application

Anti-GAPDH Antibody detects level of GAPDH & has been published & validated for use in WB.
Research Category
Metabolism
Western Blot (SNAP ID) Analysis: 0.125 µg/ml from a previous lot detected GAPDH on 10 µg of NIH/3T3 and PC12 cell lysates.

Quality

Evaluated by Western Blot in HEK293 and PC12 cell lysate.

Western Blot Analysis: 0.0625 µg/ml of this antibody detected GAPDH on 10 µg of HEK293 and PC12 cell lysate.

Target description

~ 38 kDa

Physical form

Affinity purified
Purified chicken polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to lot specific datasheet.
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transgenic APP expression during postnatal development causes persistent locomotor hyperactivity in the adult.
Rodgers, SP; Born, HA; Das, P; Jankowsky, JL
Mol. Neurodegener. null
Brian S Muntean et al.
The Journal of biological chemistry, 291(13), 7195-7204 (2016-01-27)
Regulators of G protein Signaling (RGS) promote deactivation of heterotrimeric G proteins thus controlling the magnitude and kinetics of responses mediated by G protein-coupled receptors (GPCR). In the nervous system, RGS7 and RGS9-2 play essential role in vision, reward processing
Acid-sensing ion channels contribute to synaptic transmission and inhibit cocaine-evoked plasticity.
Kreple, CJ; Lu, Y; Taugher, RJ; Schwager-Gutman, AL; Du, J; Stump, M; Wang, Y; Ghobbeh et al.
Nature Neuroscience null
Zhixiang Xu et al.
PloS one, 8(5), e63598-e63598 (2013-05-10)
Glycogen synthase kinase 3β (GSK3β), a multifaceted kinase, is abundantly expressed in the brain, including the olfactory bulb (OB). In resting cells, GSK3β is constitutively active, and its over-activation is presumably involved in numerous brain diseases, such as Alzheimer's disease.
Michael J Yetman et al.
PloS one, 11(4), e0153724-e0153724 (2016-04-14)
Alzheimer's disease (AD) researchers have struggled for decades to draw a causal link between extracellular Aβ aggregation and intraneuronal accumulation of microtubule-associated protein tau. The amyloid cascade hypothesis posits that Aβ deposition promotes tau hyperphosphorylation, tangle formation, cell loss, vascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service