Skip to Content
Merck
All Photos(1)

Key Documents

CB1001

Sigma-Aldrich

Anti-GAPDH Mouse mAb (6C5)

liquid, clone 6C5, Calbiochem®

Synonym(s):

Anti-Glyceraldehyde-3-Phosphate Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6C5, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

frog, human, porcine, chicken, canine, rabbit, mouse, fish, rat

should not react with

bovine, yeast

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)
mouse ... Gapdh(14433)
rabbit ... Gapdh(100009074)

General description

Anti-GAPDH, mouse monoclonal, clone 6C5, recognizes ~146 kDa GAPDH protein under non-reducing conditions & 36 kDa monomers under reducing conditions. It is validated for use in ELISA, WB, and IHC.
Protein A purified mouse monoclonal antibody. Recognizes the ~146 kDa GAPDH protein under non-reducing conditions and the ~36 kDa monomeric subunits under reducing conditions.
Recognizes the ~146 kDa GAPDH protein under non-reducing conditions and the ~36 kDa monomeric subunits under reducing conditions.

Immunogen

Rabbit
purified, rabbit muscle

Application


Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Highly Toxic (H)

Physical form

In PBS, pH 7.4.

Reconstitution

Following initial use, aliquot and freeze (-20°C) for long-term storage if desired. Avoid freeze/thaw.

Analysis Note

Positive Control
Rat skeletal muscle

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nastaran Masoudi-Khoram et al.
Scientific reports, 10(1), 9054-9054 (2020-06-05)
Radiotherapy is a fundamental step in the treatment of breast cancer patients. The treatment efficiency is however reduced by the possible onset of radiation resistance. In order to develop the effective treatment approach, it is important to understand molecular basis
Karina M Cirone et al.
Cell and tissue research, 379(1), 207-217 (2019-09-04)
Mycobacterium avium subsp. paratuberculosis (MAP) causes chronic diarrheic intestinal infections in domestic and wild ruminants (paratuberculosis or Johne's disease) for which there is no effective treatment. Critical in the pathogenesis of MAP infection is the invasion and survival into macrophages
Jessica L Binder et al.
PloS one, 15(3), e0230026-e0230026 (2020-03-26)
Pathological accumulation of microtubule associated protein tau in neurons is a major neuropathological hallmark of Alzheimer's disease (AD) and related tauopathies. Several attempts have been made to promote clearance of pathological tau (p-Tau) from neurons. Transcription factor EB (TFEB) has
Krizelle Mae M Alcantara et al.
Oncology reports, 41(4), 2103-2116 (2019-03-01)
Inactivation of the tumor suppressor protein Merlin leads to the development of benign nervous system tumors in neurofibromatosis type2 (NF2). Documented causes of Merlin inactivation include deleterious mutations in the encoding neurofibromin2 gene (NF2) and aberrant Merlin phosphorylation leading to
Saleh Khawaled et al.
Signal transduction and targeted therapy, 5(1), 43-43 (2020-04-18)
Tumor progression and metastasis are the major causes of death among cancer associated mortality. Metastatic cells acquire features of migration and invasion and usually undergo epithelia-mesenchymal transition (EMT). Acquirement of these various hallmarks rely on different cellular pathways, including TGF-β

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service