Skip to Content
Merck
All Photos(1)

Key Documents

U8881

Sigma-Aldrich

UCL 1684 ditrifluoroacetate hydrate

Synonym(s):

6,12,19,20,25,26-hexahydro- 5,27:13,18:21,24-Trietheno-11,7-metheno-7H-dibenzo[b,m][1,5,12,16]tetraazacyclotricosine-5,13-diium ditrifluoroacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C34H30N4)2+ · (C2F3O2-)2 · xH2O
CAS Number:
Molecular Weight:
720.66 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

color

white to off-white

solubility

DMSO: 10 mg/mL

SMILES string

[H]O[H].FC(F)(C(O)=O)F.FC(F)(C(O)=O)F.C12=CC=CC=C1C(NCC3=CC=C(CNC4=CC=[N+](C5)C6=C4C=CC=C6)C=C3)=CC=[N+]2CC7=CC=CC5=C7

InChI

1S/C34H28N4.2C3H3F3O2.H2O/c1-3-10-33-29(8-1)31-16-18-37(33)23-27-6-5-7-28(20-27)24-38-19-17-32(30-9-2-4-11-34(30)38)36-22-26-14-12-25(13-15-26)21-35-31;2*4-3(5,6)1-2(7)8;/h1-20H,21-24H2;2*1H2,(H,7,8);1H2/b35-31+,36-32+;;;

InChI key

YROHEYZVNZITJN-DDVHGCDCSA-N

Related Categories

Application

UCL 1684 ditrifluoroacetate hydrate has been used for analyzing the mechanism for ammonium-mediated displacement of the protonated analyte from analyte-trifluoroacetic acid (TFA) ion-pair. UCL 1684 has also been used as a potassium channel inhibitor to study CNP (C-type natriuretic peptide)-induced relaxations in pig coronary arteries.

Biochem/physiol Actions

UCL 1684 is a potent non-peptide blocker of the apamin-sensitive Ca2+-activated Kchannel. It comprises two quinolinium rings and is similar to apamin arginine residue. UCL 1684 inhibits calcium-activated K current (Kslow) and modulates islet β-cell.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

UCL 1684 ditrifluoroacetate hydrate is soluble in water at 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P M Dunn
European journal of pharmacology, 368(1), 119-123 (1999-03-30)
The novel K+ channel blocker 6,10-diaza-3(1,3)8,(1,4)-dibenzena-1,5(1,4)-diquinolinacy clodecaphane (UCL 1684) has been tested for its ability to inhibit Ca2+ -activated K+ currents in cultured rat chromaffin cells. Low nanomolar concentrations of UCL 1684 produced a rapid and reversible inhibition of the
Chao Fan Liang et al.
The Journal of pharmacology and experimental therapeutics, 334(1), 223-231 (2010-03-25)
The present experiments investigated whether endothelium-derived mediators modulate the effect of natriuretic peptides in porcine coronary arteries. Rings with and without endothelium were suspended in organ chambers for isometric tension recording. Concentration-relaxation curves to C-type natriuretic peptide (CNP) and atrial
Crucial role of a shared extracellular loop in apamin sensitivity and maintenance of pore shape of small-conductance calcium-activated potassium (SK) channels
Weatherall KL, et al.
Proceedings of the National Academy of Sciences of the USA, 108(45), 18494-18499 (2011)
UCL 1684: a potent blocker of Ca2+-activated K+ channels in rat adrenal chromaffin cells in culture
Dunn PM
European Journal of Pharmacology, 368(1), 119-123 (1999)
Pharmacological properties and functional role of Kslow current in mouse pancreatic beta-cells: SK channels contribute to Kslow tail current and modulate insulin secretion
Zhang M, et al.
The Journal of General Physiology, 126(4), 353-363 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service