Skip to Content
Merck
All Photos(1)

Key Documents

37291

Sigma-Aldrich

Dihydroethidium

BioReagent, suitable for fluorescence, ≥95% (HPCE)

Synonym(s):

2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine, 3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3
CAS Number:
Molecular Weight:
315.41
Beilstein:
5107482
MDL number:
UNSPSC Code:
12161505
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥95% (HPCE)

form

solid

solubility

acetonitrile: soluble
methanol: soluble

fluorescence

λex 358 nm; λem 461 nm
λex 392 nm; λem 410 nm in methanol

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CCN1C(c2ccccc2)c3cc(N)ccc3-c4ccc(N)cc14

InChI

1S/C21H21N3/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14/h3-13,21H,2,22-23H2,1H3

InChI key

XYJODUBPWNZLML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dihydroethidium (dHEt), a fluorescene probe, is used to detect the presence of reactive oxygen species (ROS) such as superoxides.
Redox indicator. Blue fluorescence until oxidized to ethidium.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Linkage

B-ring reduced analog of ethidium

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rossarin Suwanarusk et al.
The Journal of antimicrobial chemotherapy, 70(1), 124-129 (2014-08-26)
Methylene blue, once discarded due to its unsettling yet mild side effects, has now found a renewed place in the pharmacopoeia of modern medicine. The continued spread of drug-resistant Plasmodium vivax and Plasmodium falciparum has also led to a recent
Karina A Roth et al.
Journal of endodontics, 38(9), 1253-1256 (2012-08-16)
Root canal fillings are intended to prevent microbial proliferation over time in the canal after treatment. The objective of this study was to assess biofilm proliferation within the sealer-dentin interfaces of 2 methacrylate resin-based systems, self-etch (SE) and total-etch (TE)
Francisco R M Laurindo et al.
Methods in enzymology, 441, 237-260 (2008-06-17)
Assessment of low-level superoxide in nonphagocytic cells is crucial for assessing redox-dependent signaling pathways and the role of enzymes such as the NADPH oxidase complex. However, most superoxide probes present inherent limitations. Particularly, assessment of dihydroethidium (DHE) fluorescence is limited
Anna Rita Fetoni et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(9), 4011-4023 (2013-03-01)
This study addresses the relationship between cochlear oxidative damage and auditory cortical injury in a rat model of repeated noise exposure. To test the effect of increased antioxidant defenses, a water-soluble coenzyme Q10 analog (Qter) was used. We analyzed auditory
Laura Iarriccio et al.
PloS one, 10(11), e0142980-e0142980 (2015-11-17)
Dyskeratosis congenita is an inherited disease caused by mutations in genes coding for telomeric components. It was previously reported that expression of a dyskerin-derived peptide, GSE24.2, increases telomerase activity, regulates gene expression and decreases DNA damage and oxidative stress in

Questions

  1. How should stock solutions for item 37291, Dihydroethidium, be stored?

    1 answer
    1. The material should be soluble in ethanol, with dichloromethane (DCM) being a commonly used solvent for it.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service